Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 27286310-27286713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLJ21408
Entrez Gene ID
400512
Gene Name
uncharacterized LOC400512
Gene Aliases
-
UniGene
Hs.434412
Gene Chromosome Location
Chr.16: 27268205-27290468 on Build GRCh38
Transcripts
NR_037184.1

SNP Detail

SNP ID Chromosome Location
rs989066100 Chr.16: 27286475-27286475 on Build GRCh38
rs897075373 Chr.16: 27286607-27286607 on Build GRCh38
rs769807454 Chr.16: 27286604-27286604 on Build GRCh38
rs968911677 Chr.16: 27286509-27286509 on Build GRCh38
rs914751734 Chr.16: 27286487-27286487 on Build GRCh38
rs1029464646 Chr.16: 27286658-27286658 on Build GRCh38
rs200599662 Chr.16: 27286521-27286521 on Build GRCh38
rs528958723 Chr.16: 27286446-27286446 on Build GRCh38
rs764537549 Chr.16: 27286603-27286603 on Build GRCh38
rs946742392 Chr.16: 27286348-27286348 on Build GRCh38
rs8048932 Chr.16: 27286472-27286472 on Build GRCh38
rs979829096 Chr.16: 27286350-27286350 on Build GRCh38
rs186967418 Chr.16: 27286390-27286390 on Build GRCh38
rs896627256 Chr.16: 27286513-27286513 on Build GRCh38
rs955185743 Chr.16: 27286670-27286670 on Build GRCh38
rs28580502 Chr.16: 27286546-27286546 on Build GRCh38
rs746087466 Chr.16: 27286485-27286485 on Build GRCh38
rs148557013 Chr.16: 27286611-27286611 on Build GRCh38
rs944045414 Chr.16: 27286637-27286637 on Build GRCh38
rs994458894 Chr.16: 27286652-27286652 on Build GRCh38
rs1040999453 Chr.16: 27286688-27286688 on Build GRCh38
rs544615791 Chr.16: 27286432-27286432 on Build GRCh38
rs905596286 Chr.16: 27286552-27286552 on Build GRCh38
rs191458632 Chr.16: 27286391-27286391 on Build GRCh38
rs924255481 Chr.16: 27286379-27286379 on Build GRCh38
rs949797879 Chr.16: 27286531-27286531 on Build GRCh38

View Full Product Details