Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233275747-233276259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG16L1
Entrez Gene ID
55054
Gene Name
autophagy related 16 like 1
Gene Aliases
APG16L, ATG16A, ATG16L, IBD10, WDR30
UniGene
Hs.529322
Gene Chromosome Location
Chr.2: 233251571-233295674 on Build GRCh38
Transcripts
NM_001190266.1 NM_001190267.1 NM_017974.3 NM_030803.6 NM_198890.2 XM_005246082.2 XM_005246084.2 XM_005246086.2 XM_006712608.2 NR_003008.2

Gene Symbol
SCARNA5
Entrez Gene ID
677775
Gene Name
small Cajal body-specific RNA 5
Gene Aliases
U87
UniGene
Hs.663619
Gene Chromosome Location
Chr.2: 233275726-233276003 on Build GRCh38
Transcripts
NM_001190266.1 NM_001190267.1 NM_017974.3 NM_030803.6 NM_198890.2 XM_005246082.2 XM_005246084.2 XM_005246086.2 XM_006712608.2 NR_003008.2

SNP Detail

SNP ID Chromosome Location
rs775727808 Chr.2: 233275799-233275799 on Build GRCh38
rs901842364 Chr.2: 233276154-233276154 on Build GRCh38
rs773633795 Chr.2: 233276015-233276015 on Build GRCh38
rs889219733 Chr.2: 233276175-233276175 on Build GRCh38
rs765970952 Chr.2: 233275847-233275847 on Build GRCh38
rs552287165 Chr.2: 233275888-233275888 on Build GRCh38
rs530704606 Chr.2: 233276072-233276072 on Build GRCh38
rs1037056704 Chr.2: 233276142-233276142 on Build GRCh38
rs200060663 Chr.2: 233275892-233275892 on Build GRCh38
rs777069778 Chr.2: 233276036-233276036 on Build GRCh38
rs760461245 Chr.2: 233275918-233275919 on Build GRCh38
rs772169703 Chr.2: 233276007-233276007 on Build GRCh38
rs769107531 Chr.2: 233275966-233275966 on Build GRCh38
rs761061151 Chr.2: 233275846-233275846 on Build GRCh38
rs775563252 Chr.2: 233275832-233275833 on Build GRCh38
rs916744789 Chr.2: 233275800-233275800 on Build GRCh38
rs754791380 Chr.2: 233276228-233276228 on Build GRCh38
rs755336205 Chr.2: 233276046-233276046 on Build GRCh38
rs752692603 Chr.2: 233275865-233275865 on Build GRCh38
rs976915953 Chr.2: 233276001-233276001 on Build GRCh38
rs750756825 Chr.2: 233275824-233275824 on Build GRCh38
rs201458915 Chr.2: 233275867-233275867 on Build GRCh38
rs775097135 Chr.2: 233276050-233276050 on Build GRCh38
rs182555598 Chr.2: 233276020-233276020 on Build GRCh38
rs371125421 Chr.2: 233275923-233275923 on Build GRCh38
rs750430917 Chr.2: 233275960-233275961 on Build GRCh38
rs984994675 Chr.2: 233276092-233276092 on Build GRCh38
rs888200619 Chr.2: 233276182-233276182 on Build GRCh38
rs1053059000 Chr.2: 233276064-233276064 on Build GRCh38
rs949326092 Chr.2: 233276089-233276089 on Build GRCh38
rs141179324 Chr.2: 233275770-233275770 on Build GRCh38
rs940958735 Chr.2: 233276128-233276128 on Build GRCh38
rs774995875 Chr.2: 233275898-233275898 on Build GRCh38
rs1001987388 Chr.2: 233275915-233275915 on Build GRCh38
rs550897509 Chr.2: 233276192-233276192 on Build GRCh38
rs114349660 Chr.2: 233275818-233275818 on Build GRCh38
rs769936542 Chr.2: 233275795-233275795 on Build GRCh38
rs778873174 Chr.2: 233275775-233275775 on Build GRCh38
rs751840015 Chr.2: 233276024-233276026 on Build GRCh38
rs998778519 Chr.2: 233276205-233276205 on Build GRCh38
rs1046198932 Chr.2: 233276127-233276127 on Build GRCh38
rs763982952 Chr.2: 233275941-233275941 on Build GRCh38
rs767476448 Chr.2: 233275777-233275778 on Build GRCh38
rs202144988 Chr.2: 233275979-233275979 on Build GRCh38
rs1048103667 Chr.2: 233276168-233276168 on Build GRCh38
rs1003874474 Chr.2: 233276211-233276211 on Build GRCh38
rs1050489334 Chr.2: 233276169-233276169 on Build GRCh38
rs746763785 Chr.2: 233275791-233275791 on Build GRCh38
rs776217043 Chr.2: 233275928-233275928 on Build GRCh38
rs751316109 Chr.2: 233276181-233276181 on Build GRCh38
rs777702422 Chr.2: 233275877-233275877 on Build GRCh38
rs531160258 Chr.2: 233275975-233275975 on Build GRCh38
rs766308361 Chr.2: 233275903-233275904 on Build GRCh38
rs751480354 Chr.2: 233275948-233275948 on Build GRCh38
rs745599051 Chr.2: 233275972-233275972 on Build GRCh38
rs1002464205 Chr.2: 233276149-233276149 on Build GRCh38
rs541636295 Chr.2: 233275810-233275810 on Build GRCh38
rs775043473 Chr.2: 233276206-233276206 on Build GRCh38
rs934671997 Chr.2: 233275856-233275856 on Build GRCh38
rs201230365 Chr.2: 233275947-233275947 on Build GRCh38
rs528001038 Chr.2: 233275785-233275785 on Build GRCh38
rs769471887 Chr.2: 233276048-233276048 on Build GRCh38
rs532089870 Chr.2: 233275857-233275857 on Build GRCh38
rs76444894 Chr.2: 233276189-233276189 on Build GRCh38
rs977454295 Chr.2: 233276005-233276005 on Build GRCh38
rs754523890 Chr.2: 233275853-233275853 on Build GRCh38
rs909414447 Chr.2: 233276110-233276110 on Build GRCh38
rs1015820225 Chr.2: 233275949-233275949 on Build GRCh38
rs926006624 Chr.2: 233275852-233275852 on Build GRCh38
rs748049835 Chr.2: 233275776-233275776 on Build GRCh38
rs755255525 Chr.2: 233275974-233275974 on Build GRCh38
rs764115562 Chr.2: 233275944-233275945 on Build GRCh38
rs926050160 Chr.2: 233276044-233276044 on Build GRCh38
rs771923343 Chr.2: 233275783-233275783 on Build GRCh38
rs753509712 Chr.2: 233275849-233275849 on Build GRCh38
rs765137941 Chr.2: 233275851-233275851 on Build GRCh38
rs948193930 Chr.2: 233276066-233276066 on Build GRCh38
rs766568678 Chr.2: 233276023-233276023 on Build GRCh38
rs773719981 Chr.2: 233275913-233275913 on Build GRCh38
rs933332544 Chr.2: 233276163-233276163 on Build GRCh38
rs940662903 Chr.2: 233275832-233275832 on Build GRCh38
rs948247587 Chr.2: 233275806-233275806 on Build GRCh38
rs752212276 Chr.2: 233275937-233275937 on Build GRCh38
rs905003526 Chr.2: 233276135-233276135 on Build GRCh38
rs3792109 Chr.2: 233275771-233275771 on Build GRCh38
rs553662821 Chr.2: 233276212-233276212 on Build GRCh38
rs1029122806 Chr.2: 233275912-233275912 on Build GRCh38
rs758546997 Chr.2: 233276016-233276017 on Build GRCh38
rs373248719 Chr.2: 233275817-233275817 on Build GRCh38
rs1011393899 Chr.2: 233276178-233276178 on Build GRCh38
rs903047903 Chr.2: 233276189-233276189 on Build GRCh38
rs772738651 Chr.2: 233275899-233275899 on Build GRCh38
rs759178006 Chr.2: 233275936-233275936 on Build GRCh38
rs754464389 Chr.2: 233275774-233275774 on Build GRCh38
rs757050988 Chr.2: 233275886-233275886 on Build GRCh38
rs934641779 Chr.2: 233276055-233276055 on Build GRCh38
rs187253032 Chr.2: 233276021-233276021 on Build GRCh38
rs759831822 Chr.2: 233276047-233276047 on Build GRCh38
rs113262832 Chr.2: 233275938-233275938 on Build GRCh38

View Full Product Details