Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3150377-3150881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FASTKD5
Entrez Gene ID
60493
Gene Name
FAST kinase domains 5
Gene Aliases
dJ1187M17.5
UniGene
Hs.741168
Gene Chromosome Location
Chr.20: 3146519-3159886 on Build GRCh38
Transcripts
NM_021826.4 NM_001267584.1 NM_014948.3 NM_199415.2 NR_038395.1

Gene Symbol
UBOX5
Entrez Gene ID
22888
Gene Name
U-box domain containing 5
Gene Aliases
RNF37, UBCE7IP5, UIP5, hUIP5
UniGene
Hs.654646
Gene Chromosome Location
Chr.20: 3107573-3159910 on Build GRCh38
Transcripts
NM_021826.4 NM_001267584.1 NM_014948.3 NM_199415.2 NR_038395.1

Gene Symbol
UBOX5-AS1
Entrez Gene ID
100134015
Gene Name
UBOX5 antisense RNA 1
Gene Aliases
-
UniGene
Hs.129448
Gene Chromosome Location
Chr.20: 3106911-3150867 on Build GRCh38
Transcripts
NM_021826.4 NM_001267584.1 NM_014948.3 NM_199415.2 NR_038395.1

SNP Detail

SNP ID Chromosome Location
rs372467639 Chr.20: 3150648-3150648 on Build GRCh38
rs191609790 Chr.20: 3150826-3150826 on Build GRCh38
rs941110505 Chr.20: 3150760-3150760 on Build GRCh38
rs921324814 Chr.20: 3150499-3150499 on Build GRCh38
rs555693577 Chr.20: 3150716-3150716 on Build GRCh38
rs760180055 Chr.20: 3150659-3150659 on Build GRCh38
rs113875306 Chr.20: 3150841-3150842 on Build GRCh38
rs375396972 Chr.20: 3150415-3150415 on Build GRCh38
rs1020256393 Chr.20: 3150756-3150756 on Build GRCh38
rs1053059703 Chr.20: 3150637-3150637 on Build GRCh38
rs889516490 Chr.20: 3150485-3150485 on Build GRCh38
rs991375939 Chr.20: 3150568-3150568 on Build GRCh38
rs965425242 Chr.20: 3150761-3150761 on Build GRCh38
rs550156596 Chr.20: 3150511-3150511 on Build GRCh38
rs181630425 Chr.20: 3150701-3150701 on Build GRCh38
rs573891086 Chr.20: 3150736-3150736 on Build GRCh38
rs960898881 Chr.20: 3150551-3150551 on Build GRCh38
rs6051606 Chr.20: 3150515-3150515 on Build GRCh38
rs1005247472 Chr.20: 3150496-3150496 on Build GRCh38
rs998420968 Chr.20: 3150453-3150453 on Build GRCh38
rs956932955 Chr.20: 3150425-3150425 on Build GRCh38
rs988177894 Chr.20: 3150432-3150432 on Build GRCh38
rs41304822 Chr.20: 3150601-3150601 on Build GRCh38
rs923942612 Chr.20: 3150614-3150614 on Build GRCh38
rs866700365 Chr.20: 3150745-3150745 on Build GRCh38
rs901016049 Chr.20: 3150434-3150434 on Build GRCh38
rs186360702 Chr.20: 3150714-3150714 on Build GRCh38
rs1001065372 Chr.20: 3150670-3150670 on Build GRCh38
rs975536958 Chr.20: 3150483-3150483 on Build GRCh38
rs866299644 Chr.20: 3150676-3150676 on Build GRCh38
rs770801898 Chr.20: 3150703-3150703 on Build GRCh38
rs1035256527 Chr.20: 3150843-3150843 on Build GRCh38
rs935286996 Chr.20: 3150629-3150629 on Build GRCh38
rs747793943 Chr.20: 3150536-3150536 on Build GRCh38
rs2422858 Chr.20: 3150842-3150842 on Build GRCh38
rs556144640 Chr.20: 3150790-3150790 on Build GRCh38
rs756089437 Chr.20: 3150805-3150805 on Build GRCh38
rs190557117 Chr.20: 3150571-3150571 on Build GRCh38
rs916696727 Chr.20: 3150576-3150576 on Build GRCh38
rs1019590605 Chr.20: 3150439-3150439 on Build GRCh38
rs933863695 Chr.20: 3150728-3150728 on Build GRCh38
rs1028443126 Chr.20: 3150802-3150802 on Build GRCh38
rs889599102 Chr.20: 3150757-3150757 on Build GRCh38
rs908506202 Chr.20: 3150516-3150516 on Build GRCh38
rs939922722 Chr.20: 3150517-3150517 on Build GRCh38
rs1049536194 Chr.20: 3150465-3150465 on Build GRCh38
rs771608473 Chr.20: 3150599-3150599 on Build GRCh38
rs1017029737 Chr.20: 3150500-3150500 on Build GRCh38
rs543816444 Chr.20: 3150778-3150778 on Build GRCh38

View Full Product Details