Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24429385-24429877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADORA2A
Entrez Gene ID
135
Gene Name
adenosine A2a receptor
Gene Aliases
A2aR, ADORA2, RDC8
UniGene
Hs.197029
Gene Chromosome Location
Chr.22: 24423597-24442360 on Build GRCh38
Transcripts
NM_000675.5 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028484.2 NR_103546.1

Gene Symbol
ADORA2A-AS1
Entrez Gene ID
646023
Gene Name
ADORA2A antisense RNA 1
Gene Aliases
C22orf45
UniGene
Hs.632761
Gene Chromosome Location
Chr.22: 24429206-24495074 on Build GRCh38
Transcripts
NM_000675.5 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028484.2 NR_103546.1

Gene Symbol
SPECC1L-ADORA2A
Entrez Gene ID
101730217
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.474384
Gene Chromosome Location
Chr.22: 24270817-24442360 on Build GRCh38
Transcripts
NM_000675.5 NM_001278499.1 NM_001278500.1 NR_103543.1 NR_103544.1 NR_028484.2 NR_103546.1

SNP Detail

SNP ID Chromosome Location
rs953416245 Chr.22: 24429605-24429605 on Build GRCh38
rs1050471062 Chr.22: 24429812-24429812 on Build GRCh38
rs894849508 Chr.22: 24429494-24429494 on Build GRCh38
rs1002849009 Chr.22: 24429826-24429826 on Build GRCh38
rs527638176 Chr.22: 24429637-24429637 on Build GRCh38
rs924267282 Chr.22: 24429526-24429527 on Build GRCh38
rs764578616 Chr.22: 24429852-24429852 on Build GRCh38
rs573810712 Chr.22: 24429608-24429608 on Build GRCh38
rs575717744 Chr.22: 24429404-24429404 on Build GRCh38
rs878903204 Chr.22: 24429842-24429842 on Build GRCh38
rs972474335 Chr.22: 24429451-24429451 on Build GRCh38
rs1052685006 Chr.22: 24429713-24429713 on Build GRCh38
rs931461103 Chr.22: 24429780-24429780 on Build GRCh38
rs958762041 Chr.22: 24429656-24429656 on Build GRCh38
rs925208764 Chr.22: 24429695-24429695 on Build GRCh38
rs910328057 Chr.22: 24429730-24429730 on Build GRCh38
rs549827257 Chr.22: 24429820-24429820 on Build GRCh38
rs1016933688 Chr.22: 24429612-24429612 on Build GRCh38
rs773897074 Chr.22: 24429641-24429642 on Build GRCh38
rs958541723 Chr.22: 24429476-24429476 on Build GRCh38
rs1026052990 Chr.22: 24429430-24429430 on Build GRCh38
rs928849694 Chr.22: 24429638-24429638 on Build GRCh38
rs940856012 Chr.22: 24429436-24429436 on Build GRCh38
rs888838101 Chr.22: 24429544-24429544 on Build GRCh38
rs78784622 Chr.22: 24429777-24429777 on Build GRCh38
rs1047771767 Chr.22: 24429783-24429783 on Build GRCh38
rs985040506 Chr.22: 24429412-24429412 on Build GRCh38
rs987110219 Chr.22: 24429720-24429720 on Build GRCh38
rs533135350 Chr.22: 24429670-24429670 on Build GRCh38
rs559743370 Chr.22: 24429678-24429678 on Build GRCh38
rs981491564 Chr.22: 24429472-24429472 on Build GRCh38
rs778532641 Chr.22: 24429413-24429413 on Build GRCh38
rs995545026 Chr.22: 24429419-24429419 on Build GRCh38
rs1031060772 Chr.22: 24429541-24429541 on Build GRCh38
rs1006646311 Chr.22: 24429846-24429846 on Build GRCh38
rs183809787 Chr.22: 24429706-24429706 on Build GRCh38
rs997584977 Chr.22: 24429555-24429555 on Build GRCh38
rs939108871 Chr.22: 24429490-24429490 on Build GRCh38
rs865853337 Chr.22: 24429633-24429633 on Build GRCh38
rs898778385 Chr.22: 24429604-24429604 on Build GRCh38
rs962596767 Chr.22: 24429853-24429853 on Build GRCh38
rs947710358 Chr.22: 24429688-24429688 on Build GRCh38
rs770626849 Chr.22: 24429631-24429631 on Build GRCh38
rs149170930 Chr.22: 24429788-24429788 on Build GRCh38
rs1028941972 Chr.22: 24429575-24429575 on Build GRCh38
rs962205856 Chr.22: 24429634-24429634 on Build GRCh38
rs143265985 Chr.22: 24429806-24429806 on Build GRCh38
rs1007081778 Chr.22: 24429547-24429547 on Build GRCh38
rs943087355 Chr.22: 24429741-24429741 on Build GRCh38
rs544311214 Chr.22: 24429618-24429618 on Build GRCh38
rs991255427 Chr.22: 24429485-24429485 on Build GRCh38
rs2298383 Chr.22: 24429543-24429543 on Build GRCh38
rs918351602 Chr.22: 24429461-24429461 on Build GRCh38
rs772114946 Chr.22: 24429537-24429537 on Build GRCh38
rs947949598 Chr.22: 24429506-24429506 on Build GRCh38
rs928686201 Chr.22: 24429475-24429475 on Build GRCh38
rs571528247 Chr.22: 24429855-24429855 on Build GRCh38
rs746516060 Chr.22: 24429585-24429585 on Build GRCh38
rs539888982 Chr.22: 24429523-24429523 on Build GRCh38
rs1039907657 Chr.22: 24429756-24429756 on Build GRCh38
rs1035639533 Chr.22: 24429854-24429854 on Build GRCh38
rs1039878683 Chr.22: 24429592-24429592 on Build GRCh38
rs1056132042 Chr.22: 24429492-24429492 on Build GRCh38
rs111787660 Chr.22: 24429680-24429680 on Build GRCh38
rs1035751976 Chr.22: 24429641-24429641 on Build GRCh38
rs1025684683 Chr.22: 24429622-24429622 on Build GRCh38
rs903652792 Chr.22: 24429526-24429526 on Build GRCh38
rs560145123 Chr.22: 24429690-24429690 on Build GRCh38

View Full Product Details