Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86635597-86636147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA3P
Entrez Gene ID
9629
Gene Name
chloride channel accessory 3, pseudogene
Gene Aliases
CLCA3
UniGene
Hs.673847
Gene Chromosome Location
Chr.1: 86634276-86655376 on Build GRCh38
Transcripts
NR_024604.1 NR_135837.1

Gene Symbol
LOC105378828
Entrez Gene ID
105378828
Gene Name
uncharacterized LOC105378828
Gene Aliases
-
UniGene
Hs.175285
Gene Chromosome Location
Chr.1: 86571181-86704493 on Build GRCh38
Transcripts
NR_024604.1 NR_135837.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5992305 c.316G>A p.A106T Chr.1: 86635971-86635971 on Build GRCh38
6009351 c.350A>T p.Y117F Chr.1: 86636005-86636005 on Build GRCh38
6305245 c.186C>A p.Y62* Chr.1: 86635626-86635626 on Build GRCh38
7247949 c.214T>C p.Y72H Chr.1: 86635654-86635654 on Build GRCh38
536442 c.446G>A p.R149Q Chr.1: 86636101-86636101 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762202378 Chr.1: 86635960-86635960 on Build GRCh38
rs774749220 Chr.1: 86635954-86635954 on Build GRCh38
rs1012035833 Chr.1: 86636104-86636104 on Build GRCh38
rs777222773 Chr.1: 86636107-86636107 on Build GRCh38
rs911004611 Chr.1: 86635860-86635860 on Build GRCh38
rs367869112 Chr.1: 86635771-86635771 on Build GRCh38
rs764454312 Chr.1: 86635688-86635688 on Build GRCh38
rs755060034 Chr.1: 86635783-86635783 on Build GRCh38
rs372234514 Chr.1: 86635997-86635997 on Build GRCh38
rs372349247 Chr.1: 86635714-86635714 on Build GRCh38
rs770957305 Chr.1: 86635934-86635934 on Build GRCh38
rs149856130 Chr.1: 86635671-86635671 on Build GRCh38
rs986282089 Chr.1: 86635857-86635857 on Build GRCh38
rs752351323 Chr.1: 86636004-86636004 on Build GRCh38
rs957610830 Chr.1: 86635850-86635850 on Build GRCh38
rs768862219 Chr.1: 86636069-86636069 on Build GRCh38
rs144357452 Chr.1: 86635687-86635687 on Build GRCh38
rs115563507 Chr.1: 86635794-86635794 on Build GRCh38
rs997517282 Chr.1: 86636119-86636119 on Build GRCh38
rs893196119 Chr.1: 86635711-86635711 on Build GRCh38
rs765795761 Chr.1: 86635745-86635745 on Build GRCh38
rs888111770 Chr.1: 86635734-86635734 on Build GRCh38
rs369216151 Chr.1: 86636121-86636121 on Build GRCh38
rs747976178 Chr.1: 86635641-86635641 on Build GRCh38
rs1046809182 Chr.1: 86635715-86635715 on Build GRCh38
rs774661319 Chr.1: 86636072-86636072 on Build GRCh38
rs929786129 Chr.1: 86635987-86635987 on Build GRCh38
rs749436853 Chr.1: 86635942-86635942 on Build GRCh38
rs775403347 Chr.1: 86635935-86635935 on Build GRCh38
rs569309476 Chr.1: 86635672-86635672 on Build GRCh38
rs963983027 Chr.1: 86635870-86635870 on Build GRCh38
rs750899635 Chr.1: 86635731-86635731 on Build GRCh38
rs747239352 Chr.1: 86636097-86636097 on Build GRCh38
rs778673028 Chr.1: 86635628-86635628 on Build GRCh38
rs777617263 Chr.1: 86635913-86635913 on Build GRCh38
rs759616428 Chr.1: 86635667-86635667 on Build GRCh38
rs2292829 Chr.1: 86635966-86635966 on Build GRCh38
rs897086870 Chr.1: 86635654-86635654 on Build GRCh38
rs571143552 Chr.1: 86635877-86635877 on Build GRCh38
rs546311290 Chr.1: 86636025-86636025 on Build GRCh38
rs976658096 Chr.1: 86635944-86635944 on Build GRCh38
rs953568663 Chr.1: 86635852-86635852 on Build GRCh38
rs748907065 Chr.1: 86635620-86635620 on Build GRCh38
rs754593040 Chr.1: 86635762-86635762 on Build GRCh38
rs746930303 Chr.1: 86635917-86635917 on Build GRCh38
rs151201948 Chr.1: 86636110-86636110 on Build GRCh38
rs368952506 Chr.1: 86635995-86635995 on Build GRCh38
rs1056464625 Chr.1: 86635757-86635757 on Build GRCh38
rs753456880 Chr.1: 86635752-86635752 on Build GRCh38
rs1045962045 Chr.1: 86636077-86636077 on Build GRCh38
rs747471432 Chr.1: 86635940-86635943 on Build GRCh38
rs758130089 Chr.1: 86636018-86636018 on Build GRCh38
rs1034461873 Chr.1: 86635696-86635696 on Build GRCh38
rs773155657 Chr.1: 86635656-86635656 on Build GRCh38
rs768954360 Chr.1: 86635948-86635948 on Build GRCh38
rs757121694 Chr.1: 86636043-86636043 on Build GRCh38
rs369951479 Chr.1: 86636100-86636100 on Build GRCh38
rs768102425 Chr.1: 86635716-86635716 on Build GRCh38
rs139020365 Chr.1: 86635626-86635626 on Build GRCh38
rs111959913 Chr.1: 86635986-86635986 on Build GRCh38
rs748371735 Chr.1: 86636095-86636095 on Build GRCh38
rs74848396 Chr.1: 86635991-86635991 on Build GRCh38
rs749288884 Chr.1: 86636067-86636067 on Build GRCh38
rs973765645 Chr.1: 86635905-86635905 on Build GRCh38
rs113975556 Chr.1: 86635803-86635803 on Build GRCh38
rs753262166 Chr.1: 86635992-86635992 on Build GRCh38
rs78553504 Chr.1: 86636096-86636096 on Build GRCh38
rs781212897 Chr.1: 86636047-86636047 on Build GRCh38
rs755593744 Chr.1: 86635737-86635737 on Build GRCh38
rs771974350 Chr.1: 86635652-86635652 on Build GRCh38
rs201347500 Chr.1: 86635774-86635774 on Build GRCh38
rs376667458 Chr.1: 86636045-86636045 on Build GRCh38
rs929633859 Chr.1: 86635983-86635983 on Build GRCh38
rs894132254 Chr.1: 86636037-86636037 on Build GRCh38
rs142202132 Chr.1: 86636033-86636033 on Build GRCh38
rs1030582924 Chr.1: 86635836-86635836 on Build GRCh38
rs2292830 Chr.1: 86635692-86635692 on Build GRCh38
rs999386314 Chr.1: 86635800-86635800 on Build GRCh38
rs903221196 Chr.1: 86636112-86636112 on Build GRCh38
rs768008416 Chr.1: 86635964-86635964 on Build GRCh38
rs373518265 Chr.1: 86636101-86636101 on Build GRCh38

View Full Product Details