Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132108451-132108998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EP400NL
Entrez Gene ID
347918
Gene Name
EP400 N-terminal like
Gene Aliases
-
UniGene
Hs.122115
Gene Chromosome Location
Chr.12: 132084283-132126340 on Build GRCh38
Transcripts
NR_003290.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4445887 c.872C>T p.A291V Chr.12: 132108618-132108618 on Build GRCh38
6000385 c.907G>A p.A303T Chr.12: 132108653-132108653 on Build GRCh38
2001251 c.835G>C p.V279L Chr.12: 132108581-132108581 on Build GRCh38
4596576 c.902C>A p.S301Y Chr.12: 132108648-132108648 on Build GRCh38
1476346 c.902C>G p.S301C Chr.12: 132108648-132108648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943814315 Chr.12: 132108526-132108526 on Build GRCh38
rs7309728 Chr.12: 132108970-132108970 on Build GRCh38
rs936357122 Chr.12: 132108777-132108777 on Build GRCh38
rs746785611 Chr.12: 132108731-132108731 on Build GRCh38
rs75970732 Chr.12: 132108959-132108959 on Build GRCh38
rs926642276 Chr.12: 132108766-132108766 on Build GRCh38
rs939398888 Chr.12: 132108806-132108806 on Build GRCh38
rs1026347251 Chr.12: 132108898-132108898 on Build GRCh38
rs746270411 Chr.12: 132108640-132108640 on Build GRCh38
rs779021259 Chr.12: 132108551-132108551 on Build GRCh38
rs558794901 Chr.12: 132108639-132108639 on Build GRCh38
rs368347970 Chr.12: 132108576-132108576 on Build GRCh38
rs752838966 Chr.12: 132108595-132108595 on Build GRCh38
rs779151341 Chr.12: 132108542-132108545 on Build GRCh38
rs79965197 Chr.12: 132108686-132108686 on Build GRCh38
rs920897247 Chr.12: 132108765-132108765 on Build GRCh38
rs904091318 Chr.12: 132108851-132108851 on Build GRCh38
rs771681156 Chr.12: 132108675-132108675 on Build GRCh38
rs963345354 Chr.12: 132108885-132108885 on Build GRCh38
rs759374033 Chr.12: 132108723-132108723 on Build GRCh38
rs770236675 Chr.12: 132108651-132108651 on Build GRCh38
rs774105217 Chr.12: 132108652-132108652 on Build GRCh38
rs867625017 Chr.12: 132108623-132108623 on Build GRCh38
rs1036692917 Chr.12: 132108492-132108492 on Build GRCh38
rs567694981 Chr.12: 132108580-132108580 on Build GRCh38
rs747607713 Chr.12: 132108636-132108636 on Build GRCh38
rs554595200 Chr.12: 132108577-132108577 on Build GRCh38
rs544447096 Chr.12: 132108801-132108801 on Build GRCh38
rs777653552 Chr.12: 132108712-132108712 on Build GRCh38
rs892698707 Chr.12: 132108814-132108814 on Build GRCh38
rs767109981 Chr.12: 132108667-132108667 on Build GRCh38
rs142691571 Chr.12: 132108525-132108525 on Build GRCh38
rs374657159 Chr.12: 132108745-132108745 on Build GRCh38
rs1033771628 Chr.12: 132108862-132108862 on Build GRCh38
rs951322691 Chr.12: 132108928-132108928 on Build GRCh38
rs537738519 Chr.12: 132108547-132108547 on Build GRCh38
rs755859029 Chr.12: 132108631-132108631 on Build GRCh38
rs558844694 Chr.12: 132108715-132108715 on Build GRCh38
rs920574286 Chr.12: 132108500-132108500 on Build GRCh38
rs930633559 Chr.12: 132108536-132108536 on Build GRCh38
rs544576331 Chr.12: 132108760-132108760 on Build GRCh38
rs773922006 Chr.12: 132108630-132108630 on Build GRCh38
rs963374711 Chr.12: 132108692-132108692 on Build GRCh38
rs777600722 Chr.12: 132108561-132108561 on Build GRCh38
rs1002732507 Chr.12: 132108685-132108685 on Build GRCh38
rs377288238 Chr.12: 132108637-132108637 on Build GRCh38
rs982623030 Chr.12: 132108744-132108744 on Build GRCh38
rs945532551 Chr.12: 132108837-132108837 on Build GRCh38
rs753027404 Chr.12: 132108735-132108735 on Build GRCh38
rs76851765 Chr.12: 132108626-132108626 on Build GRCh38
rs747725139 Chr.12: 132108674-132108674 on Build GRCh38
rs899549996 Chr.12: 132108563-132108564 on Build GRCh38
rs553466369 Chr.12: 132108666-132108666 on Build GRCh38
rs10902497 Chr.12: 132108815-132108815 on Build GRCh38
rs1056481863 Chr.12: 132108810-132108810 on Build GRCh38
rs112427798 Chr.12: 132108476-132108476 on Build GRCh38
rs749010255 Chr.12: 132108694-132108695 on Build GRCh38
rs764085762 Chr.12: 132108886-132108886 on Build GRCh38
rs1003593727 Chr.12: 132108565-132108565 on Build GRCh38
rs560592664 Chr.12: 132108895-132108895 on Build GRCh38
rs973952461 Chr.12: 132108709-132108709 on Build GRCh38
rs189153050 Chr.12: 132108826-132108826 on Build GRCh38
rs764984411 Chr.12: 132108730-132108730 on Build GRCh38
rs940597019 Chr.12: 132108484-132108484 on Build GRCh38
rs182436721 Chr.12: 132108678-132108678 on Build GRCh38
rs1043924944 Chr.12: 132108850-132108850 on Build GRCh38
rs540600566 Chr.12: 132108894-132108894 on Build GRCh38
rs980708059 Chr.12: 132108764-132108764 on Build GRCh38
rs1046550336 Chr.12: 132108844-132108844 on Build GRCh38
rs191066353 Chr.12: 132108540-132108540 on Build GRCh38
rs983145381 Chr.12: 132108951-132108951 on Build GRCh38
rs570088614 Chr.12: 132108596-132108596 on Build GRCh38
rs1028692902 Chr.12: 132108670-132108670 on Build GRCh38

View Full Product Details