Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 103224910-103225478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPY19L2P2
Entrez Gene ID
349152
Gene Name
DPY19L2 pseudogene 2
Gene Aliases
-
UniGene
Hs.732579
Gene Chromosome Location
Chr.7: 103175013-103280466 on Build GRCh38
Transcripts
NR_003561.2 NR_027768.1

SNP Detail

SNP ID Chromosome Location
rs750182694 Chr.7: 103225125-103225125 on Build GRCh38
rs758214985 Chr.7: 103225136-103225136 on Build GRCh38
rs957394225 Chr.7: 103225332-103225332 on Build GRCh38
rs1013576847 Chr.7: 103225248-103225248 on Build GRCh38
rs1025896934 Chr.7: 103225143-103225143 on Build GRCh38
rs889532251 Chr.7: 103225043-103225043 on Build GRCh38
rs779535339 Chr.7: 103225137-103225137 on Build GRCh38
rs531217512 Chr.7: 103225402-103225402 on Build GRCh38
rs1050834862 Chr.7: 103225032-103225032 on Build GRCh38
rs771414474 Chr.7: 103225196-103225196 on Build GRCh38
rs780652822 Chr.7: 103225180-103225180 on Build GRCh38
rs767579591 Chr.7: 103225061-103225061 on Build GRCh38
rs531906105 Chr.7: 103225185-103225185 on Build GRCh38
rs560539469 Chr.7: 103225372-103225372 on Build GRCh38
rs764211701 Chr.7: 103225091-103225091 on Build GRCh38
rs969518821 Chr.7: 103225277-103225277 on Build GRCh38
rs746153962 Chr.7: 103225211-103225211 on Build GRCh38
rs758033102 Chr.7: 103225393-103225393 on Build GRCh38
rs1023748515 Chr.7: 103225250-103225250 on Build GRCh38
rs746700092 Chr.7: 103225159-103225159 on Build GRCh38
rs933655546 Chr.7: 103225029-103225029 on Build GRCh38
rs377491319 Chr.7: 103224935-103224935 on Build GRCh38
rs549420056 Chr.7: 103225451-103225451 on Build GRCh38
rs979894712 Chr.7: 103225288-103225288 on Build GRCh38
rs763745367 Chr.7: 103225167-103225167 on Build GRCh38
rs548915399 Chr.7: 103225364-103225364 on Build GRCh38
rs775811431 Chr.7: 103225082-103225082 on Build GRCh38
rs527571847 Chr.7: 103225202-103225202 on Build GRCh38
rs867978554 Chr.7: 103225129-103225129 on Build GRCh38
rs747724367 Chr.7: 103225181-103225181 on Build GRCh38
rs760620527 Chr.7: 103225090-103225090 on Build GRCh38
rs2909122 Chr.7: 103225200-103225200 on Build GRCh38
rs543367939 Chr.7: 103225457-103225457 on Build GRCh38
rs932653488 Chr.7: 103225309-103225319 on Build GRCh38
rs754009853 Chr.7: 103225094-103225094 on Build GRCh38
rs754640596 Chr.7: 103225162-103225162 on Build GRCh38
rs757251705 Chr.7: 103225109-103225109 on Build GRCh38
rs560332829 Chr.7: 103225166-103225166 on Build GRCh38
rs942887957 Chr.7: 103225047-103225047 on Build GRCh38
rs765319106 Chr.7: 103225122-103225122 on Build GRCh38
rs576097674 Chr.7: 103225139-103225139 on Build GRCh38
rs750051463 Chr.7: 103225161-103225161 on Build GRCh38

View Full Product Details