Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42943204-42943371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOOK3
Entrez Gene ID
84376
Gene Name
hook microtubule-tethering protein 3
Gene Aliases
HK3
UniGene
Hs.162852
Gene Chromosome Location
Chr.8: 42896890-43030539 on Build GRCh38
Transcripts
NM_032410.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1496897 c.279G>A p.Q93Q Chr.8: 42943324-42943324 on Build GRCh38
223252 c.320G>A p.G107E Chr.8: 42943365-42943365 on Build GRCh38
1489309 c.325C>A p.H109N Chr.8: 42943370-42943370 on Build GRCh38
1457195 c.290A>G p.D97G Chr.8: 42943335-42943335 on Build GRCh38
4646210 c.292T>A p.F98I Chr.8: 42943337-42943337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780805819 Chr.8: 42943303-42943303 on Build GRCh38
rs377365204 Chr.8: 42943277-42943288 on Build GRCh38
rs749792898 Chr.8: 42943315-42943315 on Build GRCh38
rs766189987 Chr.8: 42943290-42943290 on Build GRCh38
rs746221640 Chr.8: 42943349-42943349 on Build GRCh38
rs771534591 Chr.8: 42943320-42943320 on Build GRCh38
rs373167898 Chr.8: 42943251-42943252 on Build GRCh38
rs201764558 Chr.8: 42943293-42943293 on Build GRCh38
rs913733729 Chr.8: 42943228-42943228 on Build GRCh38
rs377741050 Chr.8: 42943252-42943257 on Build GRCh38
rs758305101 Chr.8: 42943287-42943287 on Build GRCh38
rs979184120 Chr.8: 42943245-42943245 on Build GRCh38
rs757016620 Chr.8: 42943252-42943252 on Build GRCh38
rs754670830 Chr.8: 42943294-42943294 on Build GRCh38
rs201668854 Chr.8: 42943285-42943285 on Build GRCh38
rs775183561 Chr.8: 42943327-42943327 on Build GRCh38
rs775771387 Chr.8: 42943267-42943267 on Build GRCh38
rs575405164 Chr.8: 42943282-42943282 on Build GRCh38
rs370607424 Chr.8: 42943276-42943276 on Build GRCh38
rs777290521 Chr.8: 42943307-42943307 on Build GRCh38
rs568485713 Chr.8: 42943232-42943232 on Build GRCh38
rs755596660 Chr.8: 42943306-42943306 on Build GRCh38
rs747773074 Chr.8: 42943305-42943305 on Build GRCh38

View Full Product Details