Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96200920-96201087 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLNK
Entrez Gene ID
29760
Gene Name
B-cell linker
Gene Aliases
AGM4, BASH, BLNK-S, LY57, SLP-65, SLP65, bca
UniGene
Hs.665244
Gene Chromosome Location
Chr.10: 96189171-96271576 on Build GRCh38
Transcripts
NM_001114094.1 NM_001258440.1 NM_001258441.1 NM_001258442.1 NM_013314.3 NR_047680.1 NR_047681.1 NR_047682.1 NR_047683.1 XM_011539728.2 XM_017016159.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4563221 c.963G>A p.V321V Chr.10: 96201030-96201030 on Build GRCh38
6067125 c.942A>T p.T314T Chr.10: 96201051-96201051 on Build GRCh38
6067126 c.942A>T p.T314T Chr.10: 96201051-96201051 on Build GRCh38
6873281 c.968G>T p.G323V Chr.10: 96201025-96201025 on Build GRCh38
6871114 c.959C>T p.T320I Chr.10: 96201034-96201034 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782317167 Chr.10: 96200986-96200986 on Build GRCh38
rs368853735 Chr.10: 96201035-96201035 on Build GRCh38
rs138558972 Chr.10: 96201022-96201022 on Build GRCh38
rs781904443 Chr.10: 96201045-96201045 on Build GRCh38
rs782051821 Chr.10: 96201054-96201054 on Build GRCh38
rs149590334 Chr.10: 96201036-96201036 on Build GRCh38
rs782086992 Chr.10: 96201015-96201015 on Build GRCh38
rs782713078 Chr.10: 96201058-96201058 on Build GRCh38
rs1018374824 Chr.10: 96200952-96200952 on Build GRCh38
rs587660304 Chr.10: 96200963-96200963 on Build GRCh38
rs1005955224 Chr.10: 96200942-96200942 on Build GRCh38
rs782358356 Chr.10: 96201017-96201017 on Build GRCh38
rs369093277 Chr.10: 96201024-96201024 on Build GRCh38
rs782222286 Chr.10: 96200951-96200951 on Build GRCh38
rs782646647 Chr.10: 96200962-96200962 on Build GRCh38
rs782607250 Chr.10: 96200945-96200945 on Build GRCh38
rs781940051 Chr.10: 96201014-96201014 on Build GRCh38
rs782369884 Chr.10: 96200961-96200961 on Build GRCh38

View Full Product Details