Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 142864602-142864776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB6
Entrez Gene ID
2051
Gene Name
EPH receptor B6
Gene Aliases
HEP
UniGene
Hs.380089
Gene Chromosome Location
Chr.7: 142854640-142871093 on Build GRCh38
Transcripts
NM_001280794.2 NM_001280795.2 NM_004445.5 NR_104001.2 XM_006715881.2 XM_011515879.1 XM_011515880.1 XM_011515881.2 XM_011515882.2

SNP Detail

SNP ID Chromosome Location
rs34611896 Chr.7: 142864647-142864647 on Build GRCh38
rs538788029 Chr.7: 142864627-142864627 on Build GRCh38
rs745965668 Chr.7: 142864696-142864696 on Build GRCh38
rs201782375 Chr.7: 142864665-142864665 on Build GRCh38
rs372701796 Chr.7: 142864640-142864640 on Build GRCh38
rs188945253 Chr.7: 142864756-142864756 on Build GRCh38
rs774381060 Chr.7: 142864644-142864644 on Build GRCh38
rs753188587 Chr.7: 142864680-142864680 on Build GRCh38
rs55728646 Chr.7: 142864729-142864729 on Build GRCh38
rs760406367 Chr.7: 142864754-142864754 on Build GRCh38
rs767517856 Chr.7: 142864624-142864624 on Build GRCh38
rs761705124 Chr.7: 142864721-142864721 on Build GRCh38
rs759619979 Chr.7: 142864639-142864639 on Build GRCh38
rs748119509 Chr.7: 142864642-142864643 on Build GRCh38
rs145544148 Chr.7: 142864649-142864650 on Build GRCh38
rs771280815 Chr.7: 142864692-142864692 on Build GRCh38
rs549672232 Chr.7: 142864693-142864693 on Build GRCh38
rs531094099 Chr.7: 142864664-142864664 on Build GRCh38
rs528051156 Chr.7: 142864728-142864728 on Build GRCh38
rs561585141 Chr.7: 142864701-142864701 on Build GRCh38
rs772887570 Chr.7: 142864749-142864749 on Build GRCh38
rs8177143 Chr.7: 142864648-142864648 on Build GRCh38
rs61732976 Chr.7: 142864649-142864649 on Build GRCh38
rs779429573 Chr.7: 142864667-142864667 on Build GRCh38
rs747391605 Chr.7: 142864691-142864691 on Build GRCh38
rs776913575 Chr.7: 142864633-142864633 on Build GRCh38
rs778054112 Chr.7: 142864685-142864685 on Build GRCh38
rs775608909 Chr.7: 142864708-142864708 on Build GRCh38
rs919100200 Chr.7: 142864740-142864740 on Build GRCh38

View Full Product Details