Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102024492-102024737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1R2
Entrez Gene ID
7850
Gene Name
interleukin 1 receptor type 2
Gene Aliases
CD121b, CDw121b, IL-1R-2, IL-1RT-2, IL-1RT2, IL1R2c, IL1RB
UniGene
Hs.25333
Gene Chromosome Location
Chr.2: 101991805-102028544 on Build GRCh38
Transcripts
NM_001261419.1 NM_004633.3 NR_048564.1 XM_006712734.3 XM_006712736.3 XM_011511801.2 XM_011511803.2 XM_011511804.2 XM_011511805.2 XM_011511808.2 XM_017004889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6685081 c.824C>T p.T275M Chr.2: 102024605-102024605 on Build GRCh38
5696089 c.826G>T p.A276S Chr.2: 102024607-102024607 on Build GRCh38
2980449 c.864C>T p.G288G Chr.2: 102024645-102024645 on Build GRCh38
569244 c.793G>A p.G265S Chr.2: 102024574-102024574 on Build GRCh38
3960882 c.831T>A p.N277K Chr.2: 102024612-102024612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761895256 Chr.2: 102024534-102024534 on Build GRCh38
rs758410824 Chr.2: 102024686-102024686 on Build GRCh38
rs369088604 Chr.2: 102024574-102024574 on Build GRCh38
rs748004411 Chr.2: 102024576-102024576 on Build GRCh38
rs777541472 Chr.2: 102024605-102024605 on Build GRCh38
rs549283341 Chr.2: 102024660-102024660 on Build GRCh38
rs1019515889 Chr.2: 102024514-102024515 on Build GRCh38
rs769231447 Chr.2: 102024618-102024618 on Build GRCh38
rs888449702 Chr.2: 102024685-102024685 on Build GRCh38
rs774542178 Chr.2: 102024624-102024624 on Build GRCh38
rs749704812 Chr.2: 102024615-102024615 on Build GRCh38
rs753708553 Chr.2: 102024566-102024566 on Build GRCh38
rs992644367 Chr.2: 102024604-102024604 on Build GRCh38
rs867840122 Chr.2: 102024619-102024619 on Build GRCh38
rs754830691 Chr.2: 102024638-102024638 on Build GRCh38
rs368435542 Chr.2: 102024711-102024711 on Build GRCh38
rs142547344 Chr.2: 102024551-102024551 on Build GRCh38
rs3218976 Chr.2: 102024655-102024655 on Build GRCh38
rs772421571 Chr.2: 102024658-102024658 on Build GRCh38
rs771800959 Chr.2: 102024593-102024593 on Build GRCh38
rs756986471 Chr.2: 102024648-102024648 on Build GRCh38
rs371817168 Chr.2: 102024681-102024681 on Build GRCh38
rs202144194 Chr.2: 102024689-102024689 on Build GRCh38
rs756165966 Chr.2: 102024556-102024556 on Build GRCh38
rs775181501 Chr.2: 102024710-102024710 on Build GRCh38
rs756933191 Chr.2: 102024701-102024701 on Build GRCh38
rs1007315581 Chr.2: 102024707-102024707 on Build GRCh38
rs770557266 Chr.2: 102024611-102024611 on Build GRCh38
rs551509426 Chr.2: 102024629-102024629 on Build GRCh38
rs780167021 Chr.2: 102024560-102024560 on Build GRCh38
rs754955562 Chr.2: 102024573-102024573 on Build GRCh38
rs746713889 Chr.2: 102024606-102024606 on Build GRCh38
rs930235049 Chr.2: 102024676-102024676 on Build GRCh38
rs960151581 Chr.2: 102024559-102024559 on Build GRCh38
rs147914094 Chr.2: 102024630-102024630 on Build GRCh38
rs776757109 Chr.2: 102024664-102024664 on Build GRCh38
rs141821381 Chr.2: 102024654-102024654 on Build GRCh38
rs367669711 Chr.2: 102024613-102024613 on Build GRCh38
rs759636734 Chr.2: 102024665-102024665 on Build GRCh38
rs780802660 Chr.2: 102024649-102024649 on Build GRCh38
rs765187964 Chr.2: 102024639-102024639 on Build GRCh38
rs3218975 Chr.2: 102024524-102024524 on Build GRCh38
rs978692147 Chr.2: 102024532-102024532 on Build GRCh38
rs780868934 Chr.2: 102024706-102024706 on Build GRCh38
rs377733536 Chr.2: 102024691-102024691 on Build GRCh38
rs374536258 Chr.2: 102024647-102024647 on Build GRCh38
rs776123222 Chr.2: 102024517-102024517 on Build GRCh38
rs766617649 Chr.2: 102024661-102024661 on Build GRCh38
rs751969491 Chr.2: 102024533-102024533 on Build GRCh38
rs550245933 Chr.2: 102024646-102024646 on Build GRCh38
rs377143078 Chr.2: 102024525-102024525 on Build GRCh38
rs370345051 Chr.2: 102024627-102024627 on Build GRCh38
rs745417232 Chr.2: 102024653-102024653 on Build GRCh38
rs566320548 Chr.2: 102024690-102024690 on Build GRCh38
rs765287682 Chr.2: 102024666-102024666 on Build GRCh38
rs56075526 Chr.2: 102024637-102024637 on Build GRCh38
rs150067707 Chr.2: 102024546-102024546 on Build GRCh38

View Full Product Details