Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 149579821-149580070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GINM1
Entrez Gene ID
116254
Gene Name
glycoprotein integral membrane 1
Gene Aliases
C6orf72, dJ12G14.2
UniGene
Hs.438872
Gene Chromosome Location
Chr.6: 149566392-149590931 on Build GRCh38
Transcripts
NM_138785.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7235050 c.150C>G p.L50L Chr.6: 149579932-149579932 on Build GRCh38
5512333 c.525G>A p.M175I Chr.6: 149579929-149579929 on Build GRCh38
6568740 c.198C>T p.L66L Chr.6: 149579980-149579980 on Build GRCh38
3948146 c.528C>G p.L176L Chr.6: 149579932-149579932 on Build GRCh38
5512334 c.147G>A p.M49I Chr.6: 149579929-149579929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747382393 Chr.6: 149579967-149579967 on Build GRCh38
rs982427774 Chr.6: 149579996-149580017 on Build GRCh38
rs965002666 Chr.6: 149579886-149579886 on Build GRCh38
rs775212681 Chr.6: 149579913-149579913 on Build GRCh38
rs564649407 Chr.6: 149579939-149579939 on Build GRCh38
rs768250214 Chr.6: 149579999-149579999 on Build GRCh38
rs771719024 Chr.6: 149580035-149580035 on Build GRCh38
rs148645680 Chr.6: 149579862-149579862 on Build GRCh38
rs753852891 Chr.6: 149579932-149579932 on Build GRCh38
rs956357545 Chr.6: 149579989-149579989 on Build GRCh38
rs988930993 Chr.6: 149579986-149579986 on Build GRCh38
rs753886714 Chr.6: 149579936-149579936 on Build GRCh38
rs761563128 Chr.6: 149579882-149579882 on Build GRCh38
rs34594150 Chr.6: 149579983-149579984 on Build GRCh38
rs767257320 Chr.6: 149579890-149579890 on Build GRCh38
rs549057231 Chr.6: 149580005-149580005 on Build GRCh38
rs752928540 Chr.6: 149579948-149579948 on Build GRCh38
rs373895852 Chr.6: 149579945-149579945 on Build GRCh38
rs957437135 Chr.6: 149579970-149579970 on Build GRCh38
rs566750699 Chr.6: 149579854-149579854 on Build GRCh38
rs568589964 Chr.6: 149579957-149579957 on Build GRCh38
rs773008457 Chr.6: 149579899-149579899 on Build GRCh38
rs748720822 Chr.6: 149579980-149579980 on Build GRCh38
rs116282884 Chr.6: 149580006-149580006 on Build GRCh38
rs747468426 Chr.6: 149579849-149579849 on Build GRCh38
rs1000417578 Chr.6: 149579955-149579955 on Build GRCh38
rs758714565 Chr.6: 149579952-149579952 on Build GRCh38
rs78098524 Chr.6: 149579870-149579870 on Build GRCh38
rs896003323 Chr.6: 149579879-149579879 on Build GRCh38
rs971350498 Chr.6: 149580030-149580030 on Build GRCh38
rs202099709 Chr.6: 149579880-149579880 on Build GRCh38
rs1023533034 Chr.6: 149579910-149579910 on Build GRCh38
rs375497080 Chr.6: 149579912-149579912 on Build GRCh38
rs965470501 Chr.6: 149580027-149580027 on Build GRCh38
rs781750095 Chr.6: 149579968-149579968 on Build GRCh38
rs761579365 Chr.6: 149580028-149580031 on Build GRCh38

View Full Product Details