Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 112824257-112824516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP11A
Entrez Gene ID
23250
Gene Name
ATPase phospholipid transporting 11A
Gene Aliases
ATPIH, ATPIS
UniGene
Hs.29189
Gene Chromosome Location
Chr.13: 112690034-112887168 on Build GRCh38
Transcripts
NM_015205.2 NM_032189.3 XM_005268299.4 XM_005268300.4 XM_005268301.4 XM_005268303.4 XM_005268305.4 XM_005268306.4 XM_011537480.2 XM_017020490.1 XM_017020491.1 XM_017020492.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6607074 c.791-2A>G p.? Chr.13: 112824342-112824342 on Build GRCh38
4046043 c.864C>T p.A288A Chr.13: 112824417-112824417 on Build GRCh38
1365534 c.809G>A p.G270E Chr.13: 112824362-112824362 on Build GRCh38
50396 c.839A>G p.Q280R Chr.13: 112824392-112824392 on Build GRCh38
1184008 c.806C>T p.T269M Chr.13: 112824359-112824359 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774038421 Chr.13: 112824410-112824410 on Build GRCh38
rs757402118 Chr.13: 112824446-112824446 on Build GRCh38
rs759669443 Chr.13: 112824456-112824456 on Build GRCh38
rs1011022341 Chr.13: 112824478-112824478 on Build GRCh38
rs968607091 Chr.13: 112824289-112824289 on Build GRCh38
rs370050667 Chr.13: 112824324-112824324 on Build GRCh38
rs371844988 Chr.13: 112824454-112824454 on Build GRCh38
rs755517837 Chr.13: 112824470-112824470 on Build GRCh38
rs927223196 Chr.13: 112824300-112824300 on Build GRCh38
rs779196249 Chr.13: 112824332-112824332 on Build GRCh38
rs7323586 Chr.13: 112824340-112824340 on Build GRCh38
rs146945029 Chr.13: 112824442-112824442 on Build GRCh38
rs1052314897 Chr.13: 112824349-112824349 on Build GRCh38
rs368019767 Chr.13: 112824448-112824448 on Build GRCh38
rs374668973 Chr.13: 112824393-112824393 on Build GRCh38
rs747352555 Chr.13: 112824467-112824467 on Build GRCh38
rs1009070726 Chr.13: 112824418-112824418 on Build GRCh38
rs59517603 Chr.13: 112824338-112824338 on Build GRCh38
rs781662434 Chr.13: 112824473-112824473 on Build GRCh38
rs769316766 Chr.13: 112824377-112824377 on Build GRCh38
rs376242436 Chr.13: 112824416-112824416 on Build GRCh38
rs759369113 Chr.13: 112824301-112824301 on Build GRCh38
rs199771130 Chr.13: 112824327-112824327 on Build GRCh38
rs780417257 Chr.13: 112824462-112824462 on Build GRCh38
rs375830333 Chr.13: 112824475-112824475 on Build GRCh38
rs749031798 Chr.13: 112824396-112824396 on Build GRCh38
rs34859649 Chr.13: 112824471-112824472 on Build GRCh38
rs777956162 Chr.13: 112824313-112824313 on Build GRCh38
rs980040670 Chr.13: 112824290-112824290 on Build GRCh38
rs751351709 Chr.13: 112824298-112824298 on Build GRCh38
rs141087263 Chr.13: 112824360-112824360 on Build GRCh38
rs114652675 Chr.13: 112824381-112824381 on Build GRCh38
rs533164394 Chr.13: 112824318-112824318 on Build GRCh38
rs767411997 Chr.13: 112824308-112824308 on Build GRCh38
rs752603843 Chr.13: 112824439-112824439 on Build GRCh38
rs372446027 Chr.13: 112824441-112824441 on Build GRCh38
rs377186055 Chr.13: 112824309-112824309 on Build GRCh38
rs767272882 Chr.13: 112824417-112824417 on Build GRCh38
rs112622425 Chr.13: 112824440-112824440 on Build GRCh38

View Full Product Details