Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 107710282-107710543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBX
Entrez Gene ID
56987
Gene Name
BBX, HMG-box containing
Gene Aliases
ARTC1, HBP2, HSPC339, MDS001
UniGene
Hs.124366
Gene Chromosome Location
Chr.3: 107522936-107811329 on Build GRCh38
Transcripts
NM_001142568.2 NM_001276286.1 NM_020235.6 XM_005247642.3 XM_005247643.3 XM_005247644.3 XM_011512999.2 XM_011513000.1 XM_011513001.1 XM_011513002.2 XM_011513003.2 XM_011513004.2 XM_011513005.2 XM_011513006.2 XM_011513007.2 XM_011513010.2 XM_017006881.1 XM_017006882.1 XM_017006883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3364927 c.30T>G p.H10Q Chr.3: 107710490-107710490 on Build GRCh38
445259 c.41G>C p.G14A Chr.3: 107710501-107710501 on Build GRCh38
7127750 c.83G>C p.W28S Chr.3: 107710543-107710543 on Build GRCh38
6577417 c.17G>C p.R6T Chr.3: 107710477-107710477 on Build GRCh38
3364926 c.30T>G p.H10Q Chr.3: 107710490-107710490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773501127 Chr.3: 107710480-107710480 on Build GRCh38
rs188715043 Chr.3: 107710398-107710398 on Build GRCh38
rs768390204 Chr.3: 107710499-107710501 on Build GRCh38
rs564577147 Chr.3: 107710457-107710457 on Build GRCh38
rs775339035 Chr.3: 107710428-107710429 on Build GRCh38
rs755426100 Chr.3: 107710516-107710516 on Build GRCh38
rs941936327 Chr.3: 107710358-107710358 on Build GRCh38
rs143853371 Chr.3: 107710468-107710468 on Build GRCh38
rs961016115 Chr.3: 107710314-107710314 on Build GRCh38
rs751612100 Chr.3: 107710447-107710447 on Build GRCh38
rs766765527 Chr.3: 107710492-107710492 on Build GRCh38
rs750781598 Chr.3: 107710403-107710403 on Build GRCh38
rs528150696 Chr.3: 107710485-107710485 on Build GRCh38
rs766740901 Chr.3: 107710419-107710419 on Build GRCh38
rs754358353 Chr.3: 107710420-107710420 on Build GRCh38
rs756508701 Chr.3: 107710411-107710411 on Build GRCh38
rs970511396 Chr.3: 107710317-107710317 on Build GRCh38
rs746723627 Chr.3: 107710452-107710452 on Build GRCh38
rs948566041 Chr.3: 107710362-107710362 on Build GRCh38
rs771621646 Chr.3: 107710423-107710424 on Build GRCh38
rs770744932 Chr.3: 107710454-107710454 on Build GRCh38
rs765761541 Chr.3: 107710519-107710519 on Build GRCh38
rs760866987 Chr.3: 107710486-107710486 on Build GRCh38
rs140186288 Chr.3: 107710461-107710461 on Build GRCh38
rs546841118 Chr.3: 107710470-107710470 on Build GRCh38
rs201416014 Chr.3: 107710518-107710518 on Build GRCh38
rs745424772 Chr.3: 107710418-107710418 on Build GRCh38
rs760637156 Chr.3: 107710427-107710428 on Build GRCh38
rs371658392 Chr.3: 107710456-107710456 on Build GRCh38
rs973859327 Chr.3: 107710329-107710329 on Build GRCh38
rs778368771 Chr.3: 107710435-107710435 on Build GRCh38
rs747675892 Chr.3: 107710444-107710444 on Build GRCh38
rs61731591 Chr.3: 107710474-107710474 on Build GRCh38
rs562041683 Chr.3: 107710355-107710355 on Build GRCh38
rs755667941 Chr.3: 107710395-107710395 on Build GRCh38
rs907744517 Chr.3: 107710356-107710356 on Build GRCh38
rs755475698 Chr.3: 107710421-107710421 on Build GRCh38
rs754201392 Chr.3: 107710495-107710495 on Build GRCh38
rs938583689 Chr.3: 107710458-107710458 on Build GRCh38
rs919665751 Chr.3: 107710350-107710350 on Build GRCh38

View Full Product Details