Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 159917724-159917957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRA1B
Entrez Gene ID
147
Gene Name
adrenoceptor alpha 1B
Gene Aliases
ADRA1, ALPHA1BAR
UniGene
Hs.368632
Gene Chromosome Location
Chr.5: 159916733-159989205 on Build GRCh38
Transcripts
NM_000679.3 XM_005265818.3 XM_005265819.2 XM_006714821.3 XM_011534435.1 XM_011534437.2 XM_011534438.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1065144 c.845A>G p.K282R Chr.5: 159917750-159917750 on Build GRCh38
5011544 c.916T>G p.C306G Chr.5: 159917821-159917821 on Build GRCh38
3783886 c.830G>A p.S277N Chr.5: 159917735-159917735 on Build GRCh38
6595483 c.901G>A p.G301S Chr.5: 159917806-159917806 on Build GRCh38
3853288 c.936C>A p.I312I Chr.5: 159917841-159917841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149628499 Chr.5: 159917790-159917790 on Build GRCh38
rs776807256 Chr.5: 159917903-159917905 on Build GRCh38
rs953349056 Chr.5: 159917924-159917924 on Build GRCh38
rs576264886 Chr.5: 159917904-159917904 on Build GRCh38
rs764657630 Chr.5: 159917745-159917745 on Build GRCh38
rs946284161 Chr.5: 159917898-159917898 on Build GRCh38
rs773653936 Chr.5: 159917860-159917860 on Build GRCh38
rs747013713 Chr.5: 159917827-159917844 on Build GRCh38
rs202097956 Chr.5: 159917788-159917788 on Build GRCh38
rs909092963 Chr.5: 159917931-159917931 on Build GRCh38
rs764414699 Chr.5: 159917875-159917875 on Build GRCh38
rs8192449 Chr.5: 159917886-159917886 on Build GRCh38
rs775757641 Chr.5: 159917851-159917851 on Build GRCh38
rs776642038 Chr.5: 159917864-159917864 on Build GRCh38
rs781171713 Chr.5: 159917791-159917791 on Build GRCh38
rs778325967 Chr.5: 159917772-159917772 on Build GRCh38
rs774367999 Chr.5: 159917806-159917806 on Build GRCh38
rs1041918383 Chr.5: 159917932-159917932 on Build GRCh38
rs554286300 Chr.5: 159917859-159917859 on Build GRCh38
rs758073042 Chr.5: 159917789-159917789 on Build GRCh38
rs569874100 Chr.5: 159917860-159917860 on Build GRCh38
rs562665730 Chr.5: 159917899-159917899 on Build GRCh38
rs748079207 Chr.5: 159917805-159917805 on Build GRCh38
rs201944859 Chr.5: 159917771-159917771 on Build GRCh38
rs772191822 Chr.5: 159917787-159917787 on Build GRCh38
rs576994119 Chr.5: 159917850-159917850 on Build GRCh38
rs1031620021 Chr.5: 159917849-159917849 on Build GRCh38
rs973874534 Chr.5: 159917842-159917842 on Build GRCh38
rs984762714 Chr.5: 159917928-159917928 on Build GRCh38
rs754360695 Chr.5: 159917746-159917746 on Build GRCh38
rs574447886 Chr.5: 159917765-159917765 on Build GRCh38

View Full Product Details