Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207326693-207326944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD55
Entrez Gene ID
1604
Gene Name
CD55 molecule (Cromer blood group)
Gene Aliases
CR, CROM, DAF, TC
UniGene
Hs.126517
Gene Chromosome Location
Chr.1: 207321472-207360966 on Build GRCh38
Transcripts
NM_000574.4 NM_001114752.2 NM_001300902.1 NM_001300903.1 NM_001300904.1 NR_125349.1 XM_017000467.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623827 c.580T>C p.Y194H Chr.1: 207326753-207326753 on Build GRCh38
3689325 c.648G>A p.P216P Chr.1: 207326821-207326821 on Build GRCh38
6623826 c.580T>C p.Y194H Chr.1: 207326753-207326753 on Build GRCh38
3671609 c.647C>A p.P216Q Chr.1: 207326820-207326820 on Build GRCh38
5134712 c.648G>A p.P216P Chr.1: 207326821-207326821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769500039 Chr.1: 207326886-207326886 on Build GRCh38
rs1053346766 Chr.1: 207326767-207326767 on Build GRCh38
rs150286783 Chr.1: 207326824-207326824 on Build GRCh38
rs745335968 Chr.1: 207326884-207326884 on Build GRCh38
rs778130563 Chr.1: 207326863-207326863 on Build GRCh38
rs144692928 Chr.1: 207326820-207326820 on Build GRCh38
rs776192547 Chr.1: 207326883-207326883 on Build GRCh38
rs111893615 Chr.1: 207326808-207326808 on Build GRCh38
rs755099304 Chr.1: 207326831-207326831 on Build GRCh38
rs762289167 Chr.1: 207326765-207326765 on Build GRCh38
rs764004292 Chr.1: 207326819-207326819 on Build GRCh38
rs779388813 Chr.1: 207326850-207326850 on Build GRCh38
rs146687590 Chr.1: 207326821-207326821 on Build GRCh38
rs747363356 Chr.1: 207326864-207326864 on Build GRCh38
rs533667347 Chr.1: 207326799-207326799 on Build GRCh38
rs765799348 Chr.1: 207326889-207326890 on Build GRCh38
rs1011846453 Chr.1: 207326790-207326790 on Build GRCh38
rs186881575 Chr.1: 207326890-207326890 on Build GRCh38
rs759142695 Chr.1: 207326740-207326740 on Build GRCh38
rs777025024 Chr.1: 207326727-207326728 on Build GRCh38
rs567156112 Chr.1: 207326823-207326823 on Build GRCh38
rs770565129 Chr.1: 207326876-207326876 on Build GRCh38
rs556188227 Chr.1: 207326853-207326853 on Build GRCh38
rs765247702 Chr.1: 207326770-207326770 on Build GRCh38
rs758918365 Chr.1: 207326860-207326860 on Build GRCh38
rs762832824 Chr.1: 207326807-207326807 on Build GRCh38

View Full Product Details