Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 93134253-93134526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf87
Entrez Gene ID
389676
Gene Name
chromosome 8 open reading frame 87
Gene Aliases
-
UniGene
Hs.386711
Gene Chromosome Location
Chr.8: 93134095-93166850 on Build GRCh38
Transcripts
NM_001242668.1

SNP Detail

SNP ID Chromosome Location
rs150589966 Chr.8: 93134362-93134362 on Build GRCh38
rs777700927 Chr.8: 93134497-93134497 on Build GRCh38
rs79906536 Chr.8: 93134407-93134407 on Build GRCh38
rs747958419 Chr.8: 93134448-93134448 on Build GRCh38
rs376540415 Chr.8: 93134313-93134313 on Build GRCh38
rs544863662 Chr.8: 93134361-93134361 on Build GRCh38
rs139665065 Chr.8: 93134365-93134365 on Build GRCh38
rs567084806 Chr.8: 93134415-93134415 on Build GRCh38
rs890724646 Chr.8: 93134328-93134328 on Build GRCh38
rs115051890 Chr.8: 93134387-93134387 on Build GRCh38
rs1034858058 Chr.8: 93134403-93134403 on Build GRCh38
rs765916773 Chr.8: 93134394-93134394 on Build GRCh38
rs746790701 Chr.8: 93134278-93134278 on Build GRCh38
rs560776191 Chr.8: 93134374-93134374 on Build GRCh38
rs957940280 Chr.8: 93134334-93134334 on Build GRCh38
rs13267247 Chr.8: 93134427-93134427 on Build GRCh38
rs755201653 Chr.8: 93134420-93134420 on Build GRCh38
rs937564741 Chr.8: 93134471-93134471 on Build GRCh38
rs1043084211 Chr.8: 93134293-93134293 on Build GRCh38
rs181885435 Chr.8: 93134406-93134406 on Build GRCh38
rs950591809 Chr.8: 93134396-93134396 on Build GRCh38
rs769511051 Chr.8: 93134487-93134487 on Build GRCh38
rs934798928 Chr.8: 93134308-93134308 on Build GRCh38
rs1009149365 Chr.8: 93134344-93134344 on Build GRCh38
rs190232682 Chr.8: 93134398-93134398 on Build GRCh38
rs534349447 Chr.8: 93134423-93134423 on Build GRCh38
rs754207484 Chr.8: 93134367-93134367 on Build GRCh38
rs762452945 Chr.8: 93134286-93134286 on Build GRCh38
rs926163652 Chr.8: 93134306-93134306 on Build GRCh38
rs994590489 Chr.8: 93134371-93134371 on Build GRCh38
rs970010200 Chr.8: 93134419-93134419 on Build GRCh38
rs531386579 Chr.8: 93134379-93134379 on Build GRCh38
rs772243246 Chr.8: 93134349-93134349 on Build GRCh38
rs767168312 Chr.8: 93134366-93134366 on Build GRCh38

View Full Product Details