Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114092505-114092764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF12
Entrez Gene ID
113220
Gene Name
kinesin family member 12
Gene Aliases
-
UniGene
Hs.28149
Gene Chromosome Location
Chr.9: 114091623-114100099 on Build GRCh38
Transcripts
NM_138424.1 XM_005251683.4 XM_006716947.2 XM_011518190.2 XM_011518191.2 XM_011518192.2 XM_017014240.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4462049 c.1629C>T p.P543P Chr.9: 114092595-114092595 on Build GRCh38
3779573 c.1236C>T p.P412P Chr.9: 114092589-114092589 on Build GRCh38
3779574 c.1635C>T p.P545P Chr.9: 114092589-114092589 on Build GRCh38
4626306 c.1195G>A p.E399K Chr.9: 114092630-114092630 on Build GRCh38
5128976 c.1208G>A p.G403D Chr.9: 114092617-114092617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546758730 Chr.9: 114092719-114092719 on Build GRCh38
rs144419396 Chr.9: 114092628-114092628 on Build GRCh38
rs376000496 Chr.9: 114092556-114092556 on Build GRCh38
rs748121328 Chr.9: 114092661-114092661 on Build GRCh38
rs764681898 Chr.9: 114092549-114092549 on Build GRCh38
rs140779223 Chr.9: 114092574-114092574 on Build GRCh38
rs763668900 Chr.9: 114092566-114092566 on Build GRCh38
rs780468289 Chr.9: 114092579-114092579 on Build GRCh38
rs780331338 Chr.9: 114092620-114092620 on Build GRCh38
rs754416654 Chr.9: 114092597-114092597 on Build GRCh38
rs1048793213 Chr.9: 114092600-114092600 on Build GRCh38
rs758615422 Chr.9: 114092611-114092611 on Build GRCh38
rs970480112 Chr.9: 114092686-114092686 on Build GRCh38
rs762557720 Chr.9: 114092559-114092559 on Build GRCh38
rs761643999 Chr.9: 114092687-114092690 on Build GRCh38
rs562865979 Chr.9: 114092560-114092560 on Build GRCh38
rs768464051 Chr.9: 114092636-114092636 on Build GRCh38
rs561160228 Chr.9: 114092702-114092702 on Build GRCh38
rs750645574 Chr.9: 114092610-114092610 on Build GRCh38
rs757329803 Chr.9: 114092599-114092599 on Build GRCh38
rs764636382 Chr.9: 114092582-114092582 on Build GRCh38
rs966666532 Chr.9: 114092617-114092617 on Build GRCh38
rs888252426 Chr.9: 114092640-114092640 on Build GRCh38
rs754054625 Chr.9: 114092725-114092725 on Build GRCh38
rs148915370 Chr.9: 114092704-114092704 on Build GRCh38
rs549402270 Chr.9: 114092678-114092678 on Build GRCh38
rs1023321877 Chr.9: 114092671-114092671 on Build GRCh38
rs1003259376 Chr.9: 114092708-114092708 on Build GRCh38
rs778910753 Chr.9: 114092603-114092603 on Build GRCh38
rs781071534 Chr.9: 114092646-114092646 on Build GRCh38
rs956574894 Chr.9: 114092734-114092734 on Build GRCh38
rs560252436 Chr.9: 114092543-114092543 on Build GRCh38
rs746753735 Chr.9: 114092622-114092622 on Build GRCh38

View Full Product Details