Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68678043-68678288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH3
Entrez Gene ID
1001
Gene Name
cadherin 3
Gene Aliases
CDHP, HJMD, PCAD
UniGene
Hs.191842
Gene Chromosome Location
Chr.16: 68644820-68727573 on Build GRCh38
Transcripts
NM_001317195.1 NM_001317196.1 NM_001793.5 XM_011522800.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7165776 c.378G>C p.Q126H Chr.16: 68678265-68678265 on Build GRCh38
4391809 c.370T>C p.F124L Chr.16: 68678257-68678257 on Build GRCh38
7165775 c.378G>C p.Q126H Chr.16: 68678265-68678265 on Build GRCh38
5713850 c.390G>A p.Q130Q Chr.16: 68678277-68678277 on Build GRCh38
4391808 c.370T>C p.F124L Chr.16: 68678257-68678257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760449547 Chr.16: 68678121-68678121 on Build GRCh38
rs749297263 Chr.16: 68678226-68678226 on Build GRCh38
rs182861880 Chr.16: 68678076-68678076 on Build GRCh38
rs759001232 Chr.16: 68678147-68678147 on Build GRCh38
rs138829996 Chr.16: 68678232-68678232 on Build GRCh38
rs368848411 Chr.16: 68678129-68678129 on Build GRCh38
rs375403917 Chr.16: 68678195-68678195 on Build GRCh38
rs748067951 Chr.16: 68678106-68678106 on Build GRCh38
rs764758443 Chr.16: 68678159-68678159 on Build GRCh38
rs746810978 Chr.16: 68678256-68678256 on Build GRCh38
rs1048263452 Chr.16: 68678218-68678218 on Build GRCh38
rs986076451 Chr.16: 68678084-68678084 on Build GRCh38
rs763234444 Chr.16: 68678262-68678262 on Build GRCh38
rs796911735 Chr.16: 68678140-68678140 on Build GRCh38
rs752129637 Chr.16: 68678164-68678164 on Build GRCh38
rs1056746755 Chr.16: 68678205-68678205 on Build GRCh38
rs866810651 Chr.16: 68678089-68678089 on Build GRCh38
rs774185337 Chr.16: 68678104-68678104 on Build GRCh38
rs202071102 Chr.16: 68678255-68678255 on Build GRCh38
rs149093853 Chr.16: 68678172-68678172 on Build GRCh38
rs758988304 Chr.16: 68678167-68678167 on Build GRCh38
rs771880991 Chr.16: 68678116-68678116 on Build GRCh38
rs752024803 Chr.16: 68678185-68678185 on Build GRCh38
rs374789174 Chr.16: 68678100-68678100 on Build GRCh38
rs773031095 Chr.16: 68678119-68678119 on Build GRCh38
rs766034167 Chr.16: 68678124-68678124 on Build GRCh38
rs990629693 Chr.16: 68678136-68678136 on Build GRCh38
rs757633932 Chr.16: 68678186-68678186 on Build GRCh38
rs745955311 Chr.16: 68678098-68678098 on Build GRCh38
rs369712865 Chr.16: 68678202-68678202 on Build GRCh38
rs564081886 Chr.16: 68678161-68678161 on Build GRCh38
rs780231573 Chr.16: 68678211-68678211 on Build GRCh38
rs886052229 Chr.16: 68678205-68678205 on Build GRCh38
rs755566067 Chr.16: 68678121-68678124 on Build GRCh38
rs1952048 Chr.16: 68678096-68678096 on Build GRCh38
rs373545976 Chr.16: 68678179-68678179 on Build GRCh38

View Full Product Details