Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 130379510-130379775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL6A5
Entrez Gene ID
256076
Gene Name
collagen type VI alpha 5
Gene Aliases
COL29A1, VWA4
UniGene
Hs.205403
Gene Chromosome Location
Chr.3: 130345516-130484846 on Build GRCh38
Transcripts
NM_001278298.1 NM_153264.6 NR_022012.2 XM_011512621.2 XM_011512622.2 XM_011512623.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3427044 c.860C>A p.S287Y Chr.3: 130379610-130379610 on Build GRCh38
1616878 c.1025C>A p.A342E Chr.3: 130379775-130379775 on Build GRCh38
3427043 c.817C>T p.R273* Chr.3: 130379567-130379567 on Build GRCh38
1616877 c.947C>T p.A316V Chr.3: 130379697-130379697 on Build GRCh38
4912644 c.765G>T p.Q255H Chr.3: 130379515-130379515 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966254211 Chr.3: 130379548-130379548 on Build GRCh38
rs76775308 Chr.3: 130379722-130379722 on Build GRCh38
rs13064476 Chr.3: 130379575-130379575 on Build GRCh38
rs1052693622 Chr.3: 130379629-130379629 on Build GRCh38
rs1050100877 Chr.3: 130379567-130379567 on Build GRCh38
rs548996198 Chr.3: 130379555-130379555 on Build GRCh38
rs368190316 Chr.3: 130379688-130379688 on Build GRCh38
rs751875067 Chr.3: 130379731-130379731 on Build GRCh38
rs776992312 Chr.3: 130379683-130379683 on Build GRCh38
rs764099312 Chr.3: 130379749-130379749 on Build GRCh38
rs537886317 Chr.3: 130379658-130379658 on Build GRCh38
rs558203527 Chr.3: 130379569-130379569 on Build GRCh38
rs921953985 Chr.3: 130379574-130379574 on Build GRCh38
rs1036187598 Chr.3: 130379691-130379691 on Build GRCh38
rs540827343 Chr.3: 130379626-130379626 on Build GRCh38
rs755965049 Chr.3: 130379593-130379593 on Build GRCh38
rs778150654 Chr.3: 130379614-130379614 on Build GRCh38
rs867684833 Chr.3: 130379604-130379604 on Build GRCh38
rs370536035 Chr.3: 130379543-130379543 on Build GRCh38
rs754241244 Chr.3: 130379669-130379669 on Build GRCh38
rs754497509 Chr.3: 130379651-130379651 on Build GRCh38
rs763135675 Chr.3: 130379734-130379734 on Build GRCh38
rs146891189 Chr.3: 130379625-130379625 on Build GRCh38
rs905132791 Chr.3: 130379743-130379743 on Build GRCh38
rs757739603 Chr.3: 130379671-130379671 on Build GRCh38
rs976274616 Chr.3: 130379568-130379568 on Build GRCh38
rs771403296 Chr.3: 130379680-130379680 on Build GRCh38
rs770138918 Chr.3: 130379692-130379692 on Build GRCh38

View Full Product Details