Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 102424364-102424589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL18RAP
Entrez Gene ID
8807
Gene Name
interleukin 18 receptor accessory protein
Gene Aliases
ACPL, CD218b, CDw218b, IL-18R-beta, IL-18RAcP, IL-18Rbeta, IL-1R-7, IL-1R7, IL-1RAcPL, IL18RB
UniGene
Hs.158315
Gene Chromosome Location
Chr.2: 102418551-102452568 on Build GRCh38
Transcripts
NM_003853.3 XM_011512087.2 XM_011512088.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2980768 c.564G>A p.A188A Chr.2: 102424399-102424399 on Build GRCh38
714931 c.530G>A p.S177N Chr.2: 102424365-102424365 on Build GRCh38
3564526 c.559C>T p.P187S Chr.2: 102424394-102424394 on Build GRCh38
6685048 c.569C>T p.T190I Chr.2: 102424404-102424404 on Build GRCh38
5859792 c.563C>T p.A188V Chr.2: 102424398-102424398 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190417015 Chr.2: 102424493-102424493 on Build GRCh38
rs758375788 Chr.2: 102424453-102424453 on Build GRCh38
rs762421024 Chr.2: 102424430-102424430 on Build GRCh38
rs756271882 Chr.2: 102424457-102424457 on Build GRCh38
rs766579580 Chr.2: 102424458-102424458 on Build GRCh38
rs887972 Chr.2: 102424485-102424485 on Build GRCh38
rs757177496 Chr.2: 102424444-102424444 on Build GRCh38
rs1006766100 Chr.2: 102424537-102424537 on Build GRCh38
rs754041640 Chr.2: 102424462-102424462 on Build GRCh38
rs928364392 Chr.2: 102424397-102424397 on Build GRCh38
rs139554960 Chr.2: 102424398-102424398 on Build GRCh38
rs371835232 Chr.2: 102424399-102424399 on Build GRCh38
rs759037414 Chr.2: 102424420-102424420 on Build GRCh38
rs1014935487 Chr.2: 102424492-102424492 on Build GRCh38
rs569615504 Chr.2: 102424540-102424540 on Build GRCh38
rs548191915 Chr.2: 102424535-102424535 on Build GRCh38
rs952029311 Chr.2: 102424472-102424472 on Build GRCh38
rs764682157 Chr.2: 102424425-102424425 on Build GRCh38
rs966109501 Chr.2: 102424513-102424513 on Build GRCh38
rs777459863 Chr.2: 102424411-102424411 on Build GRCh38
rs750881713 Chr.2: 102424443-102424443 on Build GRCh38
rs969563592 Chr.2: 102424395-102424395 on Build GRCh38
rs778872377 Chr.2: 102424387-102424387 on Build GRCh38
rs774930936 Chr.2: 102424428-102424428 on Build GRCh38
rs185699406 Chr.2: 102424439-102424439 on Build GRCh38
rs980257211 Chr.2: 102424396-102424396 on Build GRCh38
rs767975990 Chr.2: 102424432-102424432 on Build GRCh38
rs931198801 Chr.2: 102424445-102424445 on Build GRCh38
rs746660045 Chr.2: 102424403-102424403 on Build GRCh38
rs148604128 Chr.2: 102424404-102424404 on Build GRCh38
rs1041285906 Chr.2: 102424545-102424545 on Build GRCh38

View Full Product Details