Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 39108130-39108391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FYB
Entrez Gene ID
2533
Gene Name
FYN binding protein
Gene Aliases
ADAP, PRO0823, SLAP-130, SLAP130
UniGene
Hs.370503
Gene Chromosome Location
Chr.5: 39105252-39274523 on Build GRCh38
Transcripts
NM_001243093.1 NM_001465.4 NM_199335.3 XM_006714464.2 XM_006714465.2 XM_006714466.2 XM_011514008.2 XM_011514009.1 XM_011514010.1 XM_011514011.2 XM_017009316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7309184 c.2329+4A>T p.? Chr.5: 39108227-39108227 on Build GRCh38
5466460 c.2318A>C p.D773A Chr.5: 39108242-39108242 on Build GRCh38
5466462 c.2456A>C p.D819A Chr.5: 39108242-39108242 on Build GRCh38
7309182 c.2467+4A>T p.? Chr.5: 39108227-39108227 on Build GRCh38
3697243 c.2450A>G p.Y817C Chr.5: 39108248-39108248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758950252 Chr.5: 39108281-39108281 on Build GRCh38
rs199938361 Chr.5: 39108301-39108301 on Build GRCh38
rs911227669 Chr.5: 39108189-39108189 on Build GRCh38
rs948180771 Chr.5: 39108253-39108253 on Build GRCh38
rs768860053 Chr.5: 39108241-39108241 on Build GRCh38
rs752254544 Chr.5: 39108288-39108288 on Build GRCh38
rs377022 Chr.5: 39108207-39108207 on Build GRCh38
rs764968557 Chr.5: 39108282-39108282 on Build GRCh38
rs774841517 Chr.5: 39108248-39108248 on Build GRCh38
rs754679142 Chr.5: 39108194-39108194 on Build GRCh38
rs770094958 Chr.5: 39108217-39108217 on Build GRCh38
rs779744124 Chr.5: 39108273-39108273 on Build GRCh38
rs752755565 Chr.5: 39108338-39108338 on Build GRCh38
rs898656536 Chr.5: 39108311-39108311 on Build GRCh38
rs567648754 Chr.5: 39108219-39108219 on Build GRCh38
rs987470779 Chr.5: 39108356-39108356 on Build GRCh38
rs376541704 Chr.5: 39108300-39108300 on Build GRCh38
rs751966830 Chr.5: 39108271-39108271 on Build GRCh38
rs771150034 Chr.5: 39108214-39108214 on Build GRCh38
rs781480995 Chr.5: 39108215-39108215 on Build GRCh38
rs746879437 Chr.5: 39108209-39108209 on Build GRCh38
rs752590168 Chr.5: 39108203-39108203 on Build GRCh38
rs430630 Chr.5: 39108220-39108220 on Build GRCh38
rs894161180 Chr.5: 39108298-39108298 on Build GRCh38
rs187258674 Chr.5: 39108274-39108274 on Build GRCh38
rs994384941 Chr.5: 39108331-39108331 on Build GRCh38
rs945932684 Chr.5: 39108270-39108270 on Build GRCh38
rs1026234938 Chr.5: 39108335-39108335 on Build GRCh38
rs549151220 Chr.5: 39108216-39108216 on Build GRCh38
rs767995605 Chr.5: 39108272-39108272 on Build GRCh38
rs778403694 Chr.5: 39108200-39108200 on Build GRCh38

View Full Product Details