Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 212863652-212863899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLVCR1
Entrez Gene ID
28982
Gene Name
feline leukemia virus subgroup C cellular receptor 1
Gene Aliases
AXPC1, FLVCR, MFSD7B, PCA, PCARP
UniGene
Hs.7055
Gene Chromosome Location
Chr.1: 212858255-212899363 on Build GRCh38
Transcripts
NM_014053.3 XM_011509446.2 XM_011509447.2 XM_011509448.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1193322 c.799A>T p.T267S Chr.1: 212863785-212863785 on Build GRCh38
5733338 c.749C>T p.A250V Chr.1: 212863735-212863735 on Build GRCh38
7108482 c.792G>A p.Q264Q Chr.1: 212863778-212863778 on Build GRCh38
4968849 c.883+8A>G p.? Chr.1: 212863877-212863877 on Build GRCh38
5845405 c.785_786delAC p.Q264fs*2 Chr.1: 212863771-212863772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539066879 Chr.1: 212863844-212863844 on Build GRCh38
rs762203417 Chr.1: 212863762-212863762 on Build GRCh38
rs770284491 Chr.1: 212863840-212863840 on Build GRCh38
rs774690148 Chr.1: 212863754-212863754 on Build GRCh38
rs755239032 Chr.1: 212863821-212863821 on Build GRCh38
rs547679833 Chr.1: 212863771-212863771 on Build GRCh38
rs747001184 Chr.1: 212863834-212863834 on Build GRCh38
rs746482522 Chr.1: 212863728-212863728 on Build GRCh38
rs779345090 Chr.1: 212863822-212863822 on Build GRCh38
rs565290486 Chr.1: 212863788-212863788 on Build GRCh38
rs745446625 Chr.1: 212863863-212863863 on Build GRCh38
rs759194991 Chr.1: 212863751-212863751 on Build GRCh38
rs769490462 Chr.1: 212863752-212863752 on Build GRCh38
rs886045927 Chr.1: 212863768-212863768 on Build GRCh38
rs754753446 Chr.1: 212863685-212863685 on Build GRCh38
rs79510627 Chr.1: 212863839-212863839 on Build GRCh38
rs758264167 Chr.1: 212863829-212863829 on Build GRCh38
rs748014232 Chr.1: 212863699-212863699 on Build GRCh38
rs960237307 Chr.1: 212863753-212863753 on Build GRCh38
rs780088597 Chr.1: 212863681-212863681 on Build GRCh38
rs1051165077 Chr.1: 212863764-212863764 on Build GRCh38
rs777808317 Chr.1: 212863722-212863722 on Build GRCh38
rs750960694 Chr.1: 212863775-212863775 on Build GRCh38
rs766483912 Chr.1: 212863818-212863818 on Build GRCh38
rs367991271 Chr.1: 212863730-212863730 on Build GRCh38
rs749580355 Chr.1: 212863756-212863757 on Build GRCh38
rs529790501 Chr.1: 212863820-212863820 on Build GRCh38
rs773064101 Chr.1: 212863740-212863740 on Build GRCh38
rs1025750397 Chr.1: 212863784-212863784 on Build GRCh38
rs771901323 Chr.1: 212863713-212863713 on Build GRCh38
rs183452754 Chr.1: 212863690-212863690 on Build GRCh38
rs760628858 Chr.1: 212863807-212863807 on Build GRCh38
rs1035646599 Chr.1: 212863712-212863712 on Build GRCh38
rs891219301 Chr.1: 212863767-212863767 on Build GRCh38
rs752511404 Chr.1: 212863825-212863825 on Build GRCh38
rs777525949 Chr.1: 212863833-212863833 on Build GRCh38
rs776230830 Chr.1: 212863749-212863749 on Build GRCh38

View Full Product Details