Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63616525-63616762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HERC1
Entrez Gene ID
8925
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase family member 1
Gene Aliases
p532, p619
UniGene
Hs.210385
Gene Chromosome Location
Chr.15: 63608618-63833964 on Build GRCh38
Transcripts
NM_003922.3 XM_017022699.1 XM_017022700.1 XM_017022701.1 XM_017022702.1 XM_017022703.1 XM_017022704.1 XM_017022705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2255003 c.13777A>G p.K4593E Chr.15: 63616594-63616594 on Build GRCh38
304909 c.13717G>A p.D4573N Chr.15: 63616654-63616654 on Build GRCh38
1629625 c.13841C>T p.T4614I Chr.15: 63616530-63616530 on Build GRCh38
6846072 c.13708G>T p.A4570S Chr.15: 63616663-63616663 on Build GRCh38
963709 c.13753T>G p.L4585V Chr.15: 63616618-63616618 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781363879 Chr.15: 63616598-63616598 on Build GRCh38
rs760636290 Chr.15: 63616547-63616547 on Build GRCh38
rs748693076 Chr.15: 63616600-63616600 on Build GRCh38
rs759624813 Chr.15: 63616662-63616662 on Build GRCh38
rs779029351 Chr.15: 63616576-63616580 on Build GRCh38
rs756816701 Chr.15: 63616573-63616573 on Build GRCh38
rs761451003 Chr.15: 63616706-63616706 on Build GRCh38
rs989685077 Chr.15: 63616591-63616591 on Build GRCh38
rs753469341 Chr.15: 63616562-63616562 on Build GRCh38
rs780609445 Chr.15: 63616606-63616606 on Build GRCh38
rs775085719 Chr.15: 63616679-63616679 on Build GRCh38
rs1022869035 Chr.15: 63616609-63616609 on Build GRCh38
rs371853982 Chr.15: 63616552-63616552 on Build GRCh38
rs771918202 Chr.15: 63616668-63616668 on Build GRCh38
rs778678229 Chr.15: 63616585-63616585 on Build GRCh38
rs369953941 Chr.15: 63616559-63616559 on Build GRCh38
rs755401231 Chr.15: 63616595-63616595 on Build GRCh38
rs770351654 Chr.15: 63616607-63616607 on Build GRCh38
rs757514803 Chr.15: 63616570-63616573 on Build GRCh38
rs750014652 Chr.15: 63616594-63616594 on Build GRCh38
rs770964448 Chr.15: 63616654-63616654 on Build GRCh38
rs775372026 Chr.15: 63616615-63616615 on Build GRCh38
rs61751108 Chr.15: 63616655-63616655 on Build GRCh38
rs981424840 Chr.15: 63616649-63616649 on Build GRCh38
rs371818682 Chr.15: 63616710-63616710 on Build GRCh38
rs747525071 Chr.15: 63616610-63616616 on Build GRCh38
rs753806318 Chr.15: 63616698-63616698 on Build GRCh38
rs750113831 Chr.15: 63616731-63616731 on Build GRCh38
rs758732011 Chr.15: 63616587-63616600 on Build GRCh38
rs570192935 Chr.15: 63616642-63616642 on Build GRCh38
rs750782591 Chr.15: 63616582-63616583 on Build GRCh38
rs760544467 Chr.15: 63616686-63616686 on Build GRCh38

View Full Product Details