Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40389852-40390097 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIPK4
Entrez Gene ID
147746
Gene Name
homeodomain interacting protein kinase 4
Gene Aliases
-
UniGene
Hs.79363
Gene Chromosome Location
Chr.19: 40379271-40390221 on Build GRCh38
Transcripts
NM_144685.4 XM_006723036.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4581040 c.18G>A p.S6S Chr.19: 40389885-40389885 on Build GRCh38
3378745 c.42C>T p.I14I Chr.19: 40389861-40389861 on Build GRCh38
79972 c.33C>T p.Y11Y Chr.19: 40389870-40389870 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776355467 Chr.19: 40389901-40389901 on Build GRCh38
rs559482011 Chr.19: 40389924-40389924 on Build GRCh38
rs376400010 Chr.19: 40389951-40389951 on Build GRCh38
rs190304260 Chr.19: 40390046-40390046 on Build GRCh38
rs754177099 Chr.19: 40389933-40389933 on Build GRCh38
rs763832955 Chr.19: 40389894-40389894 on Build GRCh38
rs766692443 Chr.19: 40389916-40389916 on Build GRCh38
rs541725910 Chr.19: 40389960-40389960 on Build GRCh38
rs368704392 Chr.19: 40389885-40389885 on Build GRCh38
rs187355792 Chr.19: 40389958-40389958 on Build GRCh38
rs890175379 Chr.19: 40389974-40389975 on Build GRCh38
rs370355041 Chr.19: 40389888-40389888 on Build GRCh38
rs367863409 Chr.19: 40389909-40389909 on Build GRCh38
rs773646336 Chr.19: 40389880-40389880 on Build GRCh38
rs760883836 Chr.19: 40389886-40389886 on Build GRCh38
rs1007305068 Chr.19: 40389983-40389983 on Build GRCh38
rs200642012 Chr.19: 40389908-40389908 on Build GRCh38
rs1022495922 Chr.19: 40390017-40390017 on Build GRCh38
rs1011481470 Chr.19: 40389994-40389994 on Build GRCh38
rs374592673 Chr.19: 40389874-40389874 on Build GRCh38
rs563623358 Chr.19: 40389971-40389971 on Build GRCh38
rs906544815 Chr.19: 40389959-40389959 on Build GRCh38
rs781266305 Chr.19: 40389934-40389934 on Build GRCh38
rs756934202 Chr.19: 40390024-40390024 on Build GRCh38
rs1017216817 Chr.19: 40390007-40390007 on Build GRCh38
rs778906460 Chr.19: 40389930-40389958 on Build GRCh38
rs751177611 Chr.19: 40390019-40390019 on Build GRCh38
rs760319453 Chr.19: 40389890-40389890 on Build GRCh38
rs575075196 Chr.19: 40389972-40389972 on Build GRCh38
rs750273065 Chr.19: 40389910-40389910 on Build GRCh38
rs766365516 Chr.19: 40389918-40389918 on Build GRCh38
rs978344276 Chr.19: 40390060-40390060 on Build GRCh38
rs769993499 Chr.19: 40389878-40389878 on Build GRCh38
rs967363749 Chr.19: 40390018-40390018 on Build GRCh38
rs959509234 Chr.19: 40389982-40389982 on Build GRCh38
rs56183403 Chr.19: 40389882-40389882 on Build GRCh38
rs1033745148 Chr.19: 40389962-40389962 on Build GRCh38
rs752864489 Chr.19: 40389937-40389937 on Build GRCh38
rs372068607 Chr.19: 40389946-40389946 on Build GRCh38
rs754986667 Chr.19: 40389925-40389925 on Build GRCh38

View Full Product Details