Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 150666664-150666937 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERICH6
Entrez Gene ID
131831
Gene Name
glutamate rich 6
Gene Aliases
C3orf44, ERICH6A, FAM194A
UniGene
Hs.147128
Gene Chromosome Location
Chr.3: 150659885-150704563 on Build GRCh38
Transcripts
NM_001308234.1 NM_152394.4 XM_005247120.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6096834 c.1718C>A p.T573N Chr.3: 150666797-150666797 on Build GRCh38
479575 c.1640G>T p.G547V Chr.3: 150666875-150666875 on Build GRCh38
4821251 c.1608A>G p.K536K Chr.3: 150666907-150666907 on Build GRCh38
1039874 c.1670C>A p.S557Y Chr.3: 150666845-150666845 on Build GRCh38
236099 c.1631G>A p.R544H Chr.3: 150666884-150666884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs530849908 Chr.3: 150666866-150666866 on Build GRCh38
rs750635459 Chr.3: 150666761-150666761 on Build GRCh38
rs1041787424 Chr.3: 150666791-150666791 on Build GRCh38
rs200471791 Chr.3: 150666755-150666755 on Build GRCh38
rs756563001 Chr.3: 150666895-150666895 on Build GRCh38
rs764942627 Chr.3: 150666871-150666871 on Build GRCh38
rs748571923 Chr.3: 150666823-150666823 on Build GRCh38
rs770279340 Chr.3: 150666831-150666831 on Build GRCh38
rs372213685 Chr.3: 150666897-150666897 on Build GRCh38
rs370184261 Chr.3: 150666770-150666770 on Build GRCh38
rs748697448 Chr.3: 150666803-150666803 on Build GRCh38
rs780120705 Chr.3: 150666767-150666767 on Build GRCh38
rs150155678 Chr.3: 150666875-150666875 on Build GRCh38
rs764071357 Chr.3: 150666861-150666861 on Build GRCh38
rs758409573 Chr.3: 150666766-150666766 on Build GRCh38
rs138919004 Chr.3: 150666884-150666884 on Build GRCh38
rs781623762 Chr.3: 150666693-150666693 on Build GRCh38
rs758596449 Chr.3: 150666879-150666879 on Build GRCh38
rs140595078 Chr.3: 150666843-150666843 on Build GRCh38
rs538823944 Chr.3: 150666711-150666713 on Build GRCh38
rs111930006 Chr.3: 150666891-150666891 on Build GRCh38
rs771799576 Chr.3: 150666906-150666906 on Build GRCh38
rs924327465 Chr.3: 150666807-150666807 on Build GRCh38
rs199929972 Chr.3: 150666869-150666869 on Build GRCh38
rs373184930 Chr.3: 150666885-150666885 on Build GRCh38
rs912808282 Chr.3: 150666709-150666709 on Build GRCh38
rs748598903 Chr.3: 150666894-150666894 on Build GRCh38
rs747490564 Chr.3: 150666780-150666780 on Build GRCh38
rs760076837 Chr.3: 150666851-150666851 on Build GRCh38
rs142941844 Chr.3: 150666887-150666887 on Build GRCh38
rs11332654 Chr.3: 150666762-150666762 on Build GRCh38
rs771064630 Chr.3: 150666895-150666896 on Build GRCh38
rs774153717 Chr.3: 150666833-150666833 on Build GRCh38
rs772976071 Chr.3: 150666764-150666764 on Build GRCh38
rs148079036 Chr.3: 150666867-150666867 on Build GRCh38
rs781781666 Chr.3: 150666890-150666890 on Build GRCh38
rs143717273 Chr.3: 150666850-150666850 on Build GRCh38
rs199602127 Chr.3: 150666822-150666822 on Build GRCh38
rs749455725 Chr.3: 150666766-150666767 on Build GRCh38
rs114572143 Chr.3: 150666870-150666870 on Build GRCh38

View Full Product Details