Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 46138501-46138772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTCD
Entrez Gene ID
10841
Gene Name
formimidoyltransferase cyclodeaminase
Gene Aliases
LCHC1
UniGene
Hs.415846
Gene Chromosome Location
Chr.21: 46135981-46156481 on Build GRCh38
Transcripts
NM_001320412.1 NM_006657.2 NM_206965.1 XM_006723961.3 XM_006723962.3 XM_011529434.2 XM_011529435.2 XM_011529436.2 XM_011529437.2 XM_011529439.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6233611 c.1405G>A p.A469T Chr.21: 46138546-46138546 on Build GRCh38
4685791 c.1391C>T p.A464V Chr.21: 46138560-46138560 on Build GRCh38
3758976 c.1313C>T p.A438V Chr.21: 46138638-46138638 on Build GRCh38
6197080 c.1310C>T p.T437M Chr.21: 46138641-46138641 on Build GRCh38
4525990 c.1368G>A p.E456E Chr.21: 46138583-46138583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770320375 Chr.21: 46138683-46138683 on Build GRCh38
rs766229264 Chr.21: 46138666-46138666 on Build GRCh38
rs759922850 Chr.21: 46138597-46138597 on Build GRCh38
rs577319493 Chr.21: 46138710-46138710 on Build GRCh38
rs772615820 Chr.21: 46138662-46138662 on Build GRCh38
rs369638970 Chr.21: 46138592-46138592 on Build GRCh38
rs770268971 Chr.21: 46138654-46138654 on Build GRCh38
rs779682626 Chr.21: 46138574-46138574 on Build GRCh38
rs954091323 Chr.21: 46138653-46138653 on Build GRCh38
rs763845321 Chr.21: 46138598-46138598 on Build GRCh38
rs762727268 Chr.21: 46138561-46138561 on Build GRCh38
rs61735840 Chr.21: 46138587-46138587 on Build GRCh38
rs780933856 Chr.21: 46138526-46138526 on Build GRCh38
rs373803401 Chr.21: 46138687-46138687 on Build GRCh38
rs899798347 Chr.21: 46138739-46138739 on Build GRCh38
rs568872542 Chr.21: 46138527-46138527 on Build GRCh38
rs544253281 Chr.21: 46138580-46138580 on Build GRCh38
rs756945914 Chr.21: 46138731-46138731 on Build GRCh38
rs558891006 Chr.21: 46138699-46138699 on Build GRCh38
rs967474508 Chr.21: 46138745-46138745 on Build GRCh38
rs62214031 Chr.21: 46138677-46138677 on Build GRCh38
rs773933014 Chr.21: 46138586-46138586 on Build GRCh38
rs948347490 Chr.21: 46138607-46138607 on Build GRCh38
rs916792140 Chr.21: 46138721-46138721 on Build GRCh38
rs1005734881 Chr.21: 46138715-46138715 on Build GRCh38
rs753642045 Chr.21: 46138624-46138624 on Build GRCh38
rs749447695 Chr.21: 46138541-46138541 on Build GRCh38
rs55697096 Chr.21: 46138675-46138675 on Build GRCh38
rs775837925 Chr.21: 46138532-46138532 on Build GRCh38
rs766913256 Chr.21: 46138611-46138611 on Build GRCh38
rs576877652 Chr.21: 46138562-46138562 on Build GRCh38
rs755986536 Chr.21: 46138572-46138572 on Build GRCh38
rs750312666 Chr.21: 46138571-46138571 on Build GRCh38
rs777750537 Chr.21: 46138637-46138637 on Build GRCh38
rs753767995 Chr.21: 46138671-46138671 on Build GRCh38
rs556333001 Chr.21: 46138741-46138741 on Build GRCh38
rs376845160 Chr.21: 46138599-46138599 on Build GRCh38
rs61735841 Chr.21: 46138638-46138638 on Build GRCh38
rs749039145 Chr.21: 46138575-46138575 on Build GRCh38
rs1036750918 Chr.21: 46138743-46138743 on Build GRCh38
rs777099958 Chr.21: 46138584-46138585 on Build GRCh38
rs367805262 Chr.21: 46138641-46138641 on Build GRCh38
rs751825314 Chr.21: 46138746-46138746 on Build GRCh38
rs61735839 Chr.21: 46138542-46138542 on Build GRCh38
rs750414357 Chr.21: 46138612-46138612 on Build GRCh38
rs978258015 Chr.21: 46138591-46138591 on Build GRCh38
rs201966668 Chr.21: 46138615-46138615 on Build GRCh38
rs556952105 Chr.21: 46138522-46138522 on Build GRCh38
rs188377424 Chr.21: 46138560-46138560 on Build GRCh38
rs541835738 Chr.21: 46138596-46138596 on Build GRCh38
rs774194532 Chr.21: 46138656-46138656 on Build GRCh38
rs767156404 Chr.21: 46138661-46138661 on Build GRCh38
rs779047080 Chr.21: 46138581-46138581 on Build GRCh38
rs536554400 Chr.21: 46138531-46138531 on Build GRCh38
rs772849951 Chr.21: 46138694-46138694 on Build GRCh38
rs757284370 Chr.21: 46138523-46138523 on Build GRCh38
rs534467841 Chr.21: 46138698-46138698 on Build GRCh38
rs747065037 Chr.21: 46138679-46138679 on Build GRCh38
rs370432703 Chr.21: 46138680-46138680 on Build GRCh38
rs760008141 Chr.21: 46138660-46138662 on Build GRCh38
rs760169779 Chr.21: 46138663-46138663 on Build GRCh38
rs756761574 Chr.21: 46138564-46138564 on Build GRCh38
rs761525982 Chr.21: 46138657-46138657 on Build GRCh38
rs373426560 Chr.21: 46138633-46138633 on Build GRCh38
rs79021510 Chr.21: 46138614-46138614 on Build GRCh38
rs539095795 Chr.21: 46138521-46138521 on Build GRCh38
rs201527592 Chr.21: 46138645-46138645 on Build GRCh38
rs200283734 Chr.21: 46138593-46138593 on Build GRCh38
rs60525628 Chr.21: 46138672-46138672 on Build GRCh38
rs779168035 Chr.21: 46138631-46138631 on Build GRCh38
rs199807194 Chr.21: 46138543-46138543 on Build GRCh38
rs781255845 Chr.21: 46138568-46138568 on Build GRCh38
rs367649158 Chr.21: 46138695-46138695 on Build GRCh38
rs377363402 Chr.21: 46138634-46138634 on Build GRCh38
rs536289303 Chr.21: 46138676-46138676 on Build GRCh38
rs1039935726 Chr.21: 46138732-46138732 on Build GRCh38
rs368782658 Chr.21: 46138563-46138563 on Build GRCh38
rs574566550 Chr.21: 46138742-46138742 on Build GRCh38
rs771633489 Chr.21: 46138690-46138690 on Build GRCh38
rs1047179 Chr.21: 46138559-46138559 on Build GRCh38
rs894147280 Chr.21: 46138605-46138605 on Build GRCh38
rs761059490 Chr.21: 46138590-46138590 on Build GRCh38
rs544813975 Chr.21: 46138717-46138717 on Build GRCh38
rs773590831 Chr.21: 46138550-46138550 on Build GRCh38
rs376572763 Chr.21: 46138588-46138588 on Build GRCh38
rs754699274 Chr.21: 46138626-46138626 on Build GRCh38
rs374170977 Chr.21: 46138546-46138546 on Build GRCh38
rs765377200 Chr.21: 46138681-46138682 on Build GRCh38
rs373471933 Chr.21: 46138644-46138644 on Build GRCh38
rs764988572 Chr.21: 46138673-46138673 on Build GRCh38
rs761465271 Chr.21: 46138609-46138609 on Build GRCh38
rs766238466 Chr.21: 46138727-46138727 on Build GRCh38

View Full Product Details