Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 41541980-41542249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GINS4
Entrez Gene ID
84296
Gene Name
GINS complex subunit 4
Gene Aliases
SLD5
UniGene
Hs.656996
Gene Chromosome Location
Chr.8: 41529206-41545046 on Build GRCh38
Transcripts
NM_032336.2 XM_005273659.3 NR_125824.1

Gene Symbol
LOC102723729
Entrez Gene ID
102723729
Gene Name
uncharacterized LOC102723729
Gene Aliases
-
UniGene
Hs.586384
Gene Chromosome Location
Chr.8: 41534305-41578368 on Build GRCh38
Transcripts
NM_032336.2 XM_005273659.3 NR_125824.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6454155 c.583G>A p.V195M Chr.8: 41541998-41541998 on Build GRCh38
3900156 c.582C>T p.Y194Y Chr.8: 41541997-41541997 on Build GRCh38
6561477 c.589G>A p.D197N Chr.8: 41542004-41542004 on Build GRCh38
4643255 c.615G>T p.L205F Chr.8: 41542030-41542030 on Build GRCh38
7141638 c.620G>T p.R207L Chr.8: 41542035-41542035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989681676 Chr.8: 41542184-41542184 on Build GRCh38
rs371762239 Chr.8: 41542108-41542108 on Build GRCh38
rs1037180272 Chr.8: 41542193-41542193 on Build GRCh38
rs769636118 Chr.8: 41542027-41542027 on Build GRCh38
rs559630833 Chr.8: 41542189-41542189 on Build GRCh38
rs1049796376 Chr.8: 41542155-41542155 on Build GRCh38
rs758356866 Chr.8: 41542163-41542163 on Build GRCh38
rs528229647 Chr.8: 41542071-41542071 on Build GRCh38
rs544781957 Chr.8: 41542211-41542211 on Build GRCh38
rs958180209 Chr.8: 41542171-41542171 on Build GRCh38
rs779145522 Chr.8: 41542115-41542115 on Build GRCh38
rs769905897 Chr.8: 41542190-41542190 on Build GRCh38
rs552035072 Chr.8: 41542180-41542180 on Build GRCh38
rs1055455430 Chr.8: 41542007-41542007 on Build GRCh38
rs772826423 Chr.8: 41542033-41542033 on Build GRCh38
rs747997128 Chr.8: 41542025-41542025 on Build GRCh38
rs566816336 Chr.8: 41542154-41542154 on Build GRCh38
rs897320543 Chr.8: 41542196-41542196 on Build GRCh38
rs531039634 Chr.8: 41542053-41542053 on Build GRCh38
rs144574209 Chr.8: 41542072-41542072 on Build GRCh38
rs1023934224 Chr.8: 41542057-41542057 on Build GRCh38
rs764530667 Chr.8: 41542107-41542107 on Build GRCh38
rs141417166 Chr.8: 41542018-41542018 on Build GRCh38
rs746068571 Chr.8: 41542117-41542117 on Build GRCh38
rs753274950 Chr.8: 41542067-41542067 on Build GRCh38
rs564157693 Chr.8: 41542150-41542150 on Build GRCh38
rs779932169 Chr.8: 41542124-41542124 on Build GRCh38
rs757612756 Chr.8: 41542110-41542110 on Build GRCh38
rs758494264 Chr.8: 41542119-41542119 on Build GRCh38
rs774118928 Chr.8: 41542040-41542040 on Build GRCh38
rs769679898 Chr.8: 41542138-41542138 on Build GRCh38
rs779090249 Chr.8: 41542015-41542015 on Build GRCh38
rs772107093 Chr.8: 41542021-41542021 on Build GRCh38
rs912670265 Chr.8: 41542179-41542179 on Build GRCh38
rs978650862 Chr.8: 41542149-41542149 on Build GRCh38
rs759171041 Chr.8: 41542045-41542045 on Build GRCh38
rs748130894 Chr.8: 41542128-41542128 on Build GRCh38
rs546011448 Chr.8: 41542035-41542035 on Build GRCh38
rs750671145 Chr.8: 41542046-41542046 on Build GRCh38
rs766205000 Chr.8: 41542039-41542039 on Build GRCh38
rs776678766 Chr.8: 41542024-41542024 on Build GRCh38

View Full Product Details