Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 1967797-1968066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCP1B
Entrez Gene ID
196513
Gene Name
decapping mRNA 1B
Gene Aliases
DCP1
UniGene
Hs.130934
Gene Chromosome Location
Chr.12: 1941591-2004535 on Build GRCh38
Transcripts
NM_152640.4 NR_135060.1 XM_011520927.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3935870 c.323C>G p.S108C Chr.12: 1967907-1967907 on Build GRCh38
5812669 c.58A>T p.R20* Chr.12: 1967866-1967866 on Build GRCh38
4909045 c.17C>G p.S6C Chr.12: 1967907-1967907 on Build GRCh38
6640975 c.14-5delT p.? Chr.12: 1967915-1967915 on Build GRCh38
5812668 c.364A>T p.R122* Chr.12: 1967866-1967866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112977950 Chr.12: 1967919-1967919 on Build GRCh38
rs577649881 Chr.12: 1967878-1967878 on Build GRCh38
rs774244937 Chr.12: 1967834-1967834 on Build GRCh38
rs888147141 Chr.12: 1967984-1967984 on Build GRCh38
rs1019390136 Chr.12: 1968025-1968025 on Build GRCh38
rs763868741 Chr.12: 1967882-1967882 on Build GRCh38
rs889681418 Chr.12: 1967979-1967979 on Build GRCh38
rs1028254441 Chr.12: 1967939-1967939 on Build GRCh38
rs934436518 Chr.12: 1967900-1967900 on Build GRCh38
rs1008128631 Chr.12: 1968015-1968015 on Build GRCh38
rs556012795 Chr.12: 1967843-1967843 on Build GRCh38
rs760941939 Chr.12: 1967879-1967879 on Build GRCh38
rs374581892 Chr.12: 1967854-1967854 on Build GRCh38
rs778935605 Chr.12: 1967901-1967901 on Build GRCh38
rs373593739 Chr.12: 1967894-1967894 on Build GRCh38
rs753704287 Chr.12: 1967884-1967884 on Build GRCh38
rs749683287 Chr.12: 1967962-1967962 on Build GRCh38
rs749865234 Chr.12: 1967906-1967906 on Build GRCh38
rs77559005 Chr.12: 1967932-1967932 on Build GRCh38
rs890032053 Chr.12: 1967824-1967824 on Build GRCh38
rs537645798 Chr.12: 1967818-1967818 on Build GRCh38
rs879145647 Chr.12: 1967959-1967959 on Build GRCh38
rs1001598797 Chr.12: 1967999-1967999 on Build GRCh38
rs778253657 Chr.12: 1967941-1967941 on Build GRCh38
rs779796104 Chr.12: 1967931-1967931 on Build GRCh38
rs759691765 Chr.12: 1967851-1967851 on Build GRCh38

View Full Product Details