Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 21652387-21652650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGSF6
Entrez Gene ID
10261
Gene Name
immunoglobulin superfamily member 6
Gene Aliases
DORA
UniGene
Hs.530902
Gene Chromosome Location
Chr.16: 21641284-21652666 on Build GRCh38
Transcripts
NM_005849.3 NM_001077180.2 NM_001288659.1 NM_001288660.1 NM_016025.4 XM_017023269.1

Gene Symbol
METTL9
Entrez Gene ID
51108
Gene Name
methyltransferase like 9
Gene Aliases
CGI-81, DREV, DREV1, PAP1
UniGene
Hs.279583
Gene Chromosome Location
Chr.16: 21597221-21657473 on Build GRCh38
Transcripts
NM_005849.3 NM_001077180.2 NM_001288659.1 NM_001288660.1 NM_016025.4 XM_017023269.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4405515 c.25A>G p.I9V Chr.16: 21652574-21652574 on Build GRCh38
4059214 c.28G>A p.A10T Chr.16: 21652571-21652571 on Build GRCh38
5850444 c.67G>A p.G23S Chr.16: 21652532-21652532 on Build GRCh38
3948369 c.44C>G p.T15S Chr.16: 21652555-21652555 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758654423 Chr.16: 21652625-21652625 on Build GRCh38
rs565354015 Chr.16: 21652588-21652588 on Build GRCh38
rs769214915 Chr.16: 21652575-21652575 on Build GRCh38
rs3091401 Chr.16: 21652462-21652462 on Build GRCh38
rs1001735121 Chr.16: 21652428-21652428 on Build GRCh38
rs781488951 Chr.16: 21652581-21652581 on Build GRCh38
rs942618641 Chr.16: 21652599-21652599 on Build GRCh38
rs776240570 Chr.16: 21652498-21652498 on Build GRCh38
rs762741600 Chr.16: 21652605-21652605 on Build GRCh38
rs372070506 Chr.16: 21652612-21652612 on Build GRCh38
rs943644897 Chr.16: 21652536-21652536 on Build GRCh38
rs1006036389 Chr.16: 21652623-21652623 on Build GRCh38
rs142541478 Chr.16: 21652537-21652537 on Build GRCh38
rs747997806 Chr.16: 21652555-21652555 on Build GRCh38
rs758599428 Chr.16: 21652568-21652568 on Build GRCh38
rs200550835 Chr.16: 21652591-21652591 on Build GRCh38
rs960754586 Chr.16: 21652419-21652419 on Build GRCh38
rs761165783 Chr.16: 21652505-21652505 on Build GRCh38
rs143387566 Chr.16: 21652532-21652532 on Build GRCh38
rs3091402 Chr.16: 21652474-21652474 on Build GRCh38
rs760631627 Chr.16: 21652618-21652618 on Build GRCh38
rs749223817 Chr.16: 21652609-21652610 on Build GRCh38
rs910785304 Chr.16: 21652533-21652533 on Build GRCh38
rs753956942 Chr.16: 21652549-21652549 on Build GRCh38
rs767330123 Chr.16: 21652616-21652616 on Build GRCh38
rs367923647 Chr.16: 21652620-21652620 on Build GRCh38
rs980046400 Chr.16: 21652483-21652483 on Build GRCh38
rs368774015 Chr.16: 21652567-21652567 on Build GRCh38
rs376663815 Chr.16: 21652546-21652546 on Build GRCh38
rs747337935 Chr.16: 21652572-21652572 on Build GRCh38
rs750605979 Chr.16: 21652431-21652431 on Build GRCh38
rs375567163 Chr.16: 21652523-21652523 on Build GRCh38
rs544399390 Chr.16: 21652506-21652506 on Build GRCh38
rs757339391 Chr.16: 21652552-21652552 on Build GRCh38
rs989872454 Chr.16: 21652500-21652500 on Build GRCh38
rs759337843 Chr.16: 21652615-21652615 on Build GRCh38
rs772745383 Chr.16: 21652603-21652603 on Build GRCh38
rs772795315 Chr.16: 21652512-21652512 on Build GRCh38
rs554246887 Chr.16: 21652611-21652611 on Build GRCh38
rs752644777 Chr.16: 21652617-21652617 on Build GRCh38
rs3091403 Chr.16: 21652542-21652542 on Build GRCh38
rs780421744 Chr.16: 21652571-21652571 on Build GRCh38
rs576015170 Chr.16: 21652543-21652543 on Build GRCh38
rs773112804 Chr.16: 21652505-21652507 on Build GRCh38

View Full Product Details