Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78223254-78223509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC5
Entrez Gene ID
332
Gene Name
baculoviral IAP repeat containing 5
Gene Aliases
API4, EPR-1
UniGene
Hs.744872
Gene Chromosome Location
Chr.17: 78214196-78225635 on Build GRCh38
Transcripts
NM_001012270.1 NM_001012271.1 NM_001168.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1133510 c.415G>A p.E139K Chr.17: 78223471-78223471 on Build GRCh38
1680115 c.707C>A p.S236Y Chr.17: 78223492-78223492 on Build GRCh38
1680114 c.249G>T p.K83N Chr.17: 78223492-78223492 on Build GRCh38
1680113 c.367G>T p.E123* Chr.17: 78223492-78223492 on Build GRCh38
1133511 c.228G>A p.R76R Chr.17: 78223471-78223471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368810457 Chr.17: 78223420-78223420 on Build GRCh38
rs752456543 Chr.17: 78223428-78223428 on Build GRCh38
rs989986139 Chr.17: 78223340-78223340 on Build GRCh38
rs537206590 Chr.17: 78223359-78223359 on Build GRCh38
rs753320749 Chr.17: 78223480-78223480 on Build GRCh38
rs577440700 Chr.17: 78223419-78223419 on Build GRCh38
rs910493223 Chr.17: 78223394-78223394 on Build GRCh38
rs924463537 Chr.17: 78223290-78223290 on Build GRCh38
rs1006806849 Chr.17: 78223335-78223335 on Build GRCh38
rs75349024 Chr.17: 78223442-78223442 on Build GRCh38
rs142521887 Chr.17: 78223301-78223301 on Build GRCh38
rs760563717 Chr.17: 78223437-78223437 on Build GRCh38
rs565973721 Chr.17: 78223397-78223397 on Build GRCh38
rs557536118 Chr.17: 78223396-78223396 on Build GRCh38

View Full Product Details