Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 51215101-51215344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP19A1
Entrez Gene ID
1588
Gene Name
cytochrome P450 family 19 subfamily A member 1
Gene Aliases
ARO, ARO1, CPV1, CYAR, CYP19, CYPXIX, P-450AROM
UniGene
Hs.260074
Gene Chromosome Location
Chr.15: 51208057-51338598 on Build GRCh38
Transcripts
NM_000103.3 NM_031226.2 XM_005254191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
962678 c.882G>T p.E294D Chr.15: 51215209-51215209 on Build GRCh38
3502049 c.912G>A p.L304L Chr.15: 51215179-51215179 on Build GRCh38
962679 c.859-1G>A p.? Chr.15: 51215233-51215233 on Build GRCh38
7293839 c.896G>A p.C299Y Chr.15: 51215195-51215195 on Build GRCh38
317208 c.976C>T p.P326S Chr.15: 51215115-51215115 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778448927 Chr.15: 51215160-51215160 on Build GRCh38
rs370257485 Chr.15: 51215172-51215172 on Build GRCh38
rs774286243 Chr.15: 51215250-51215250 on Build GRCh38
rs758055562 Chr.15: 51215258-51215258 on Build GRCh38
rs754344873 Chr.15: 51215234-51215234 on Build GRCh38
rs765722947 Chr.15: 51215244-51215244 on Build GRCh38
rs745573360 Chr.15: 51215248-51215248 on Build GRCh38
rs371448768 Chr.15: 51215167-51215167 on Build GRCh38
rs372742123 Chr.15: 51215193-51215193 on Build GRCh38
rs1019317190 Chr.15: 51215187-51215187 on Build GRCh38
rs776512401 Chr.15: 51215190-51215190 on Build GRCh38
rs745465006 Chr.15: 51215163-51215163 on Build GRCh38
rs756838912 Chr.15: 51215238-51215238 on Build GRCh38
rs746837828 Chr.15: 51215283-51215283 on Build GRCh38
rs763044967 Chr.15: 51215191-51215191 on Build GRCh38
rs148169419 Chr.15: 51215206-51215206 on Build GRCh38
rs903468994 Chr.15: 51215159-51215159 on Build GRCh38
rs143839949 Chr.15: 51215138-51215138 on Build GRCh38
rs756723709 Chr.15: 51215228-51215228 on Build GRCh38
rs747006734 Chr.15: 51215175-51215175 on Build GRCh38
rs527734817 Chr.15: 51215294-51215294 on Build GRCh38
rs771766414 Chr.15: 51215165-51215165 on Build GRCh38
rs767831661 Chr.15: 51215213-51215213 on Build GRCh38
rs181766689 Chr.15: 51215229-51215229 on Build GRCh38
rs759803650 Chr.15: 51215208-51215208 on Build GRCh38
rs768558411 Chr.15: 51215176-51215176 on Build GRCh38
rs59359360 Chr.15: 51215128-51215128 on Build GRCh38
rs779762427 Chr.15: 51215274-51215274 on Build GRCh38
rs753093862 Chr.15: 51215225-51215225 on Build GRCh38
rs996782674 Chr.15: 51215198-51215198 on Build GRCh38
rs2289105 Chr.15: 51215311-51215311 on Build GRCh38
rs756679571 Chr.15: 51215134-51215134 on Build GRCh38
rs756783943 Chr.15: 51215139-51215139 on Build GRCh38

View Full Product Details