Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 14880090-14880347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2R1
Entrez Gene ID
120227
Gene Name
cytochrome P450 family 2 subfamily R member 1
Gene Aliases
-
UniGene
Hs.371427
Gene Chromosome Location
Chr.11: 14877436-14898913 on Build GRCh38
Transcripts
NM_024514.4 XM_005252788.1 XM_005252789.2 XM_011519895.1 XM_011519898.2 XM_017017190.1 XM_017017191.1 XM_017017192.1 XM_017017193.1 XM_017017194.1 XM_017017195.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
925344 c.958C>T p.R320W Chr.11: 14880178-14880178 on Build GRCh38
1249592 c.919C>T p.L307F Chr.11: 14880217-14880217 on Build GRCh38
925343 c.970C>A p.L324I Chr.11: 14880166-14880166 on Build GRCh38
4973344 c.913G>A p.G305S Chr.11: 14880223-14880223 on Build GRCh38
122464 c.806A>T p.K269M Chr.11: 14880330-14880330 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782483896 Chr.11: 14880163-14880163 on Build GRCh38
rs782063388 Chr.11: 14880145-14880145 on Build GRCh38
rs983042802 Chr.11: 14880282-14880282 on Build GRCh38
rs782701012 Chr.11: 14880170-14880170 on Build GRCh38
rs969616871 Chr.11: 14880173-14880173 on Build GRCh38
rs148817258 Chr.11: 14880276-14880276 on Build GRCh38
rs782770722 Chr.11: 14880161-14880161 on Build GRCh38
rs1022332516 Chr.11: 14880156-14880156 on Build GRCh38
rs782489426 Chr.11: 14880308-14880308 on Build GRCh38
rs149753792 Chr.11: 14880224-14880224 on Build GRCh38
rs782784464 Chr.11: 14880147-14880147 on Build GRCh38
rs781971106 Chr.11: 14880135-14880135 on Build GRCh38
rs782503732 Chr.11: 14880284-14880284 on Build GRCh38
rs145638214 Chr.11: 14880251-14880251 on Build GRCh38
rs782571220 Chr.11: 14880311-14880311 on Build GRCh38
rs147626987 Chr.11: 14880186-14880186 on Build GRCh38
rs781827684 Chr.11: 14880286-14880286 on Build GRCh38
rs782520338 Chr.11: 14880178-14880178 on Build GRCh38
rs782078269 Chr.11: 14880256-14880256 on Build GRCh38
rs368317393 Chr.11: 14880141-14880141 on Build GRCh38
rs782603852 Chr.11: 14880181-14880181 on Build GRCh38
rs200127496 Chr.11: 14880320-14880320 on Build GRCh38
rs781856242 Chr.11: 14880258-14880258 on Build GRCh38
rs61742603 Chr.11: 14880315-14880315 on Build GRCh38
rs145224817 Chr.11: 14880162-14880162 on Build GRCh38
rs201185910 Chr.11: 14880176-14880176 on Build GRCh38
rs34910346 Chr.11: 14880121-14880122 on Build GRCh38
rs558701830 Chr.11: 14880148-14880148 on Build GRCh38
rs782250434 Chr.11: 14880177-14880177 on Build GRCh38
rs140870202 Chr.11: 14880219-14880219 on Build GRCh38
rs552653924 Chr.11: 14880166-14880166 on Build GRCh38
rs782290266 Chr.11: 14880226-14880226 on Build GRCh38
rs200183599 Chr.11: 14880285-14880285 on Build GRCh38
rs184549196 Chr.11: 14880263-14880263 on Build GRCh38
rs201596523 Chr.11: 14880160-14880160 on Build GRCh38
rs781917192 Chr.11: 14880260-14880260 on Build GRCh38
rs367701971 Chr.11: 14880307-14880307 on Build GRCh38
rs527423165 Chr.11: 14880246-14880246 on Build GRCh38
rs781810701 Chr.11: 14880171-14880171 on Build GRCh38
rs782116417 Chr.11: 14880155-14880155 on Build GRCh38
rs782792716 Chr.11: 14880266-14880266 on Build GRCh38
rs782323196 Chr.11: 14880187-14880187 on Build GRCh38
rs1037491274 Chr.11: 14880290-14880290 on Build GRCh38
rs782346995 Chr.11: 14880128-14880128 on Build GRCh38

View Full Product Details