Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6281149-6281384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT7
Entrez Gene ID
11332
Gene Name
acyl-CoA thioesterase 7
Gene Aliases
ACH1, ACT, BACH, CTE-II, LACH, LACH1, hBACH
UniGene
Hs.126137
Gene Chromosome Location
Chr.1: 6264272-6393766 on Build GRCh38
Transcripts
NM_007274.3 NM_181864.2 NM_181865.2 NM_181866.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4511733 c.788C>T p.S263L Chr.1: 6281268-6281268 on Build GRCh38
1584662 c.726G>A p.S242S Chr.1: 6281267-6281267 on Build GRCh38
6592302 c.888C>T p.I296I Chr.1: 6281228-6281228 on Build GRCh38
6592304 c.765C>T p.I255I Chr.1: 6281228-6281228 on Build GRCh38
4956783 c.900G>A p.V300V Chr.1: 6281216-6281216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376626576 Chr.1: 6281210-6281210 on Build GRCh38
rs370084810 Chr.1: 6281310-6281343 on Build GRCh38
rs887202228 Chr.1: 6281312-6281312 on Build GRCh38
rs779960902 Chr.1: 6281319-6281319 on Build GRCh38
rs375753921 Chr.1: 6281310-6281310 on Build GRCh38
rs370806018 Chr.1: 6281316-6281316 on Build GRCh38
rs376115546 Chr.1: 6281282-6281282 on Build GRCh38
rs754831608 Chr.1: 6281224-6281224 on Build GRCh38
rs772753567 Chr.1: 6281213-6281213 on Build GRCh38
rs775787329 Chr.1: 6281193-6281193 on Build GRCh38
rs939733397 Chr.1: 6281352-6281352 on Build GRCh38
rs757434983 Chr.1: 6281174-6281174 on Build GRCh38
rs200575720 Chr.1: 6281308-6281308 on Build GRCh38
rs770045340 Chr.1: 6281182-6281182 on Build GRCh38
rs754104045 Chr.1: 6281322-6281322 on Build GRCh38
rs755203981 Chr.1: 6281337-6281337 on Build GRCh38
rs570582538 Chr.1: 6281267-6281267 on Build GRCh38
rs376229263 Chr.1: 6281338-6281338 on Build GRCh38
rs772662485 Chr.1: 6281272-6281272 on Build GRCh38
rs191303476 Chr.1: 6281309-6281309 on Build GRCh38
rs751397081 Chr.1: 6281317-6281317 on Build GRCh38
rs959354776 Chr.1: 6281235-6281235 on Build GRCh38
rs766912861 Chr.1: 6281260-6281260 on Build GRCh38
rs774864324 Chr.1: 6281209-6281209 on Build GRCh38
rs140066536 Chr.1: 6281228-6281228 on Build GRCh38
rs144257022 Chr.1: 6281301-6281334 on Build GRCh38
rs138220668 Chr.1: 6281263-6281263 on Build GRCh38
rs760299701 Chr.1: 6281216-6281216 on Build GRCh38
rs768954851 Chr.1: 6281207-6281207 on Build GRCh38
rs373628657 Chr.1: 6281256-6281256 on Build GRCh38
rs781390925 Chr.1: 6281178-6281178 on Build GRCh38
rs749670729 Chr.1: 6281201-6281201 on Build GRCh38
rs756242866 Chr.1: 6281318-6281318 on Build GRCh38
rs368731455 Chr.1: 6281212-6281212 on Build GRCh38
rs987126579 Chr.1: 6281231-6281231 on Build GRCh38
rs749887902 Chr.1: 6281304-6281304 on Build GRCh38
rs552618053 Chr.1: 6281251-6281251 on Build GRCh38
rs751964976 Chr.1: 6281318-6281319 on Build GRCh38
rs772551653 Chr.1: 6281268-6281268 on Build GRCh38
rs766047954 Chr.1: 6281219-6281219 on Build GRCh38
rs749424998 Chr.1: 6281261-6281261 on Build GRCh38
rs750203670 Chr.1: 6281171-6281171 on Build GRCh38
rs376071703 Chr.1: 6281244-6281244 on Build GRCh38
rs760234992 Chr.1: 6281281-6281281 on Build GRCh38
rs752711896 Chr.1: 6281239-6281239 on Build GRCh38
rs746127296 Chr.1: 6281181-6281181 on Build GRCh38
rs554306005 Chr.1: 6281330-6281330 on Build GRCh38
rs368769818 Chr.1: 6281305-6281305 on Build GRCh38
rs779303400 Chr.1: 6281331-6281331 on Build GRCh38
rs753364120 Chr.1: 6281221-6281221 on Build GRCh38
rs772354346 Chr.1: 6281335-6281335 on Build GRCh38
rs376035990 Chr.1: 6281315-6281315 on Build GRCh38
rs369724108 Chr.1: 6281249-6281249 on Build GRCh38
rs906710628 Chr.1: 6281220-6281220 on Build GRCh38
rs757492849 Chr.1: 6281322-6281322 on Build GRCh38

View Full Product Details