Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 15623166-15623415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC171
Entrez Gene ID
203238
Gene Name
coiled-coil domain containing 171
Gene Aliases
C9orf93, bA536D16.1, bA778P13.1
UniGene
Hs.17267
Gene Chromosome Location
Chr.9: 15552874-16062431 on Build GRCh38
Transcripts
NM_173550.2 XM_005251397.2 XM_005251398.3 XM_005251399.4 XM_011517788.2 XM_011517791.2 XM_011517796.2 XM_017014431.1 XM_017014432.1 XM_017014433.1 XM_017014434.1 XM_017014435.1 XM_017014436.1 XM_017014437.1 XM_017014438.1 XM_017014439.1 XM_017014440.1 XM_017014441.1 XM_017014442.1 XM_017014443.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1461547 c.720A>C p.K240N Chr.9: 15623311-15623311 on Build GRCh38
3090625 c.706A>G p.K236E Chr.9: 15623297-15623297 on Build GRCh38
1461548 c.752delT p.L252fs*27 Chr.9: 15623343-15623343 on Build GRCh38
7321192 c.774A>C p.E258D Chr.9: 15623365-15623365 on Build GRCh38
361314 c.767C>G p.T256R Chr.9: 15623358-15623358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543165680 Chr.9: 15623213-15623213 on Build GRCh38
rs886461143 Chr.9: 15623346-15623346 on Build GRCh38
rs759975543 Chr.9: 15623269-15623269 on Build GRCh38
rs919853685 Chr.9: 15623326-15623326 on Build GRCh38
rs201103188 Chr.9: 15623220-15623220 on Build GRCh38
rs767579990 Chr.9: 15623342-15623342 on Build GRCh38
rs569754263 Chr.9: 15623352-15623352 on Build GRCh38
rs762496830 Chr.9: 15623246-15623246 on Build GRCh38
rs761431259 Chr.9: 15623259-15623259 on Build GRCh38
rs369967566 Chr.9: 15623338-15623338 on Build GRCh38
rs956987158 Chr.9: 15623204-15623204 on Build GRCh38
rs749306503 Chr.9: 15623298-15623298 on Build GRCh38
rs769816676 Chr.9: 15623239-15623239 on Build GRCh38
rs768286724 Chr.9: 15623253-15623253 on Build GRCh38
rs770126567 Chr.9: 15623373-15623373 on Build GRCh38
rs970242220 Chr.9: 15623195-15623195 on Build GRCh38
rs202011112 Chr.9: 15623351-15623351 on Build GRCh38
rs112320377 Chr.9: 15623379-15623379 on Build GRCh38
rs747560011 Chr.9: 15623361-15623361 on Build GRCh38
rs763505675 Chr.9: 15623332-15623332 on Build GRCh38
rs771623782 Chr.9: 15623366-15623366 on Build GRCh38
rs1001705580 Chr.9: 15623377-15623377 on Build GRCh38
rs938029239 Chr.9: 15623219-15623219 on Build GRCh38
rs1047720026 Chr.9: 15623334-15623334 on Build GRCh38
rs746727594 Chr.9: 15623371-15623371 on Build GRCh38
rs754193297 Chr.9: 15623264-15623264 on Build GRCh38
rs780903016 Chr.9: 15623235-15623235 on Build GRCh38
rs774049935 Chr.9: 15623254-15623254 on Build GRCh38
rs950943828 Chr.9: 15623381-15623381 on Build GRCh38
rs752823080 Chr.9: 15623247-15623248 on Build GRCh38
rs189533353 Chr.9: 15623384-15623384 on Build GRCh38
rs759834009 Chr.9: 15623323-15623323 on Build GRCh38
rs747196228 Chr.9: 15623225-15623225 on Build GRCh38
rs754692204 Chr.9: 15623309-15623309 on Build GRCh38
rs764463714 Chr.9: 15623385-15623385 on Build GRCh38
rs1033279638 Chr.9: 15623200-15623200 on Build GRCh38
rs775593669 Chr.9: 15623240-15623240 on Build GRCh38
rs1014327388 Chr.9: 15623349-15623349 on Build GRCh38
rs750664166 Chr.9: 15623348-15623348 on Build GRCh38
rs368595197 Chr.9: 15623273-15623273 on Build GRCh38
rs955132985 Chr.9: 15623227-15623227 on Build GRCh38
rs777891810 Chr.9: 15623275-15623275 on Build GRCh38
rs751789315 Chr.9: 15623277-15623277 on Build GRCh38
rs751558158 Chr.9: 15623337-15623337 on Build GRCh38
rs143279568 Chr.9: 15623295-15623295 on Build GRCh38
rs752731681 Chr.9: 15623216-15623216 on Build GRCh38
rs544956922 Chr.9: 15623215-15623215 on Build GRCh38
rs772938660 Chr.9: 15623320-15623320 on Build GRCh38
rs184996816 Chr.9: 15623223-15623223 on Build GRCh38
rs745604019 Chr.9: 15623290-15623290 on Build GRCh38
rs201321568 Chr.9: 15623304-15623304 on Build GRCh38
rs772670746 Chr.9: 15623369-15623369 on Build GRCh38
rs770751092 Chr.9: 15623230-15623230 on Build GRCh38
rs768268159 Chr.9: 15623303-15623303 on Build GRCh38
rs765588024 Chr.9: 15623328-15623328 on Build GRCh38
rs373672355 Chr.9: 15623367-15623367 on Build GRCh38
rs866234690 Chr.9: 15623378-15623378 on Build GRCh38
rs747805540 Chr.9: 15623305-15623305 on Build GRCh38
rs766743327 Chr.9: 15623260-15623260 on Build GRCh38
rs1001758103 Chr.9: 15623198-15623198 on Build GRCh38
rs775878363 Chr.9: 15623382-15623382 on Build GRCh38
rs200870154 Chr.9: 15623340-15623340 on Build GRCh38
rs778939983 Chr.9: 15623358-15623358 on Build GRCh38
rs765682018 Chr.9: 15623270-15623270 on Build GRCh38
rs757464121 Chr.9: 15623281-15623281 on Build GRCh38
rs781512201 Chr.9: 15623285-15623285 on Build GRCh38

View Full Product Details