Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 2885280-2885541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHNO1
Entrez Gene ID
83695
Gene Name
RAD9-HUS1-RAD1 interacting nuclear orphan 1
Gene Aliases
C12orf32, HKMT1188, RHINO
UniGene
Hs.198853
Gene Chromosome Location
Chr.12: 2876258-2889525 on Build GRCh38
Transcripts
NM_001252499.2 NM_001252500.2 NM_001257097.1 NM_001257098.1 NR_027365.2 NR_046432.1 NR_046433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5840848 c.42_43CC>TT p.P15S Chr.12: 2885408-2885409 on Build GRCh38
3739596 c.68T>C p.L23P Chr.12: 2885434-2885434 on Build GRCh38
938615 c.62A>T p.Q21L Chr.12: 2885428-2885428 on Build GRCh38
5659914 c.131C>T p.P44L Chr.12: 2885497-2885497 on Build GRCh38
7426303 c.35A>T p.Q12L Chr.12: 2885401-2885401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769900816 Chr.12: 2885338-2885347 on Build GRCh38
rs564888666 Chr.12: 2885513-2885513 on Build GRCh38
rs1027029101 Chr.12: 2885326-2885326 on Build GRCh38
rs766367326 Chr.12: 2885366-2885366 on Build GRCh38
rs763382048 Chr.12: 2885463-2885463 on Build GRCh38
rs889933719 Chr.12: 2885451-2885451 on Build GRCh38
rs752859007 Chr.12: 2885475-2885475 on Build GRCh38
rs73040528 Chr.12: 2885339-2885339 on Build GRCh38
rs376284597 Chr.12: 2885362-2885362 on Build GRCh38
rs144376511 Chr.12: 2885469-2885469 on Build GRCh38
rs547329725 Chr.12: 2885467-2885467 on Build GRCh38
rs137966722 Chr.12: 2885368-2885368 on Build GRCh38
rs140138359 Chr.12: 2885479-2885479 on Build GRCh38
rs34096285 Chr.12: 2885388-2885388 on Build GRCh38
rs370802415 Chr.12: 2885360-2885360 on Build GRCh38
rs770713695 Chr.12: 2885435-2885435 on Build GRCh38
rs767087540 Chr.12: 2885385-2885385 on Build GRCh38
rs777387543 Chr.12: 2885428-2885428 on Build GRCh38
rs771295789 Chr.12: 2885447-2885462 on Build GRCh38
rs749457421 Chr.12: 2885438-2885438 on Build GRCh38
rs766990128 Chr.12: 2885464-2885464 on Build GRCh38
rs377569803 Chr.12: 2885507-2885507 on Build GRCh38
rs377262236 Chr.12: 2885465-2885465 on Build GRCh38
rs142328102 Chr.12: 2885446-2885446 on Build GRCh38
rs763830545 Chr.12: 2885460-2885460 on Build GRCh38
rs150099344 Chr.12: 2885411-2885411 on Build GRCh38
rs868163932 Chr.12: 2885425-2885425 on Build GRCh38
rs914940746 Chr.12: 2885431-2885431 on Build GRCh38
rs745853086 Chr.12: 2885495-2885495 on Build GRCh38
rs752412686 Chr.12: 2885386-2885386 on Build GRCh38
rs770822789 Chr.12: 2885487-2885487 on Build GRCh38
rs746419451 Chr.12: 2885317-2885317 on Build GRCh38
rs374668211 Chr.12: 2885459-2885459 on Build GRCh38
rs187710658 Chr.12: 2885352-2885352 on Build GRCh38
rs552720705 Chr.12: 2885384-2885384 on Build GRCh38
rs773369846 Chr.12: 2885347-2885348 on Build GRCh38
rs751624888 Chr.12: 2885402-2885402 on Build GRCh38
rs747108635 Chr.12: 2885333-2885333 on Build GRCh38
rs758481589 Chr.12: 2885329-2885329 on Build GRCh38
rs138375075 Chr.12: 2885412-2885412 on Build GRCh38
rs748966358 Chr.12: 2885485-2885485 on Build GRCh38
rs774273779 Chr.12: 2885436-2885436 on Build GRCh38
rs774661733 Chr.12: 2885481-2885482 on Build GRCh38
rs765570307 Chr.12: 2885389-2885389 on Build GRCh38
rs933137524 Chr.12: 2885516-2885516 on Build GRCh38
rs778355470 Chr.12: 2885340-2885341 on Build GRCh38
rs897230880 Chr.12: 2885477-2885477 on Build GRCh38
rs778065198 Chr.12: 2885481-2885481 on Build GRCh38
rs781466636 Chr.12: 2885474-2885474 on Build GRCh38
rs771626818 Chr.12: 2885453-2885453 on Build GRCh38
rs774939901 Chr.12: 2885509-2885509 on Build GRCh38
rs755178003 Chr.12: 2885410-2885410 on Build GRCh38
rs771539166 Chr.12: 2885505-2885505 on Build GRCh38
rs376359977 Chr.12: 2885334-2885334 on Build GRCh38
rs564361449 Chr.12: 2885438-2885438 on Build GRCh38
rs758783091 Chr.12: 2885392-2885392 on Build GRCh38
rs540252783 Chr.12: 2885488-2885488 on Build GRCh38
rs147662161 Chr.12: 2885433-2885433 on Build GRCh38
rs371402573 Chr.12: 2885396-2885396 on Build GRCh38
rs750695454 Chr.12: 2885461-2885461 on Build GRCh38
rs779958767 Chr.12: 2885330-2885330 on Build GRCh38
rs779146587 Chr.12: 2885311-2885311 on Build GRCh38

View Full Product Details