Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41444191-41444416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5SL
Entrez Gene ID
55101
Gene Name
ATP5S like
Gene Aliases
-
UniGene
Hs.351099
Gene Chromosome Location
Chr.19: 41431318-41447822 on Build GRCh38
Transcripts
XM_011527065.2 NM_001130514.1 XM_017026119.1

Gene Symbol
ERICH4
Entrez Gene ID
100170765
Gene Name
glutamate rich 4
Gene Aliases
C19orf69
UniGene
Hs.649452
Gene Chromosome Location
Chr.19: 41442426-41444765 on Build GRCh38
Transcripts
XM_011527065.2 NM_001130514.1 XM_017026119.1

SNP Detail

SNP ID Chromosome Location
rs116061667 Chr.19: 41444304-41444304 on Build GRCh38
rs765074669 Chr.19: 41444294-41444294 on Build GRCh38
rs923311408 Chr.19: 41444241-41444241 on Build GRCh38
rs560077124 Chr.19: 41444285-41444285 on Build GRCh38
rs782174108 Chr.19: 41444225-41444225 on Build GRCh38
rs976478703 Chr.19: 41444230-41444230 on Build GRCh38
rs941175448 Chr.19: 41444354-41444354 on Build GRCh38
rs782085682 Chr.19: 41444260-41444260 on Build GRCh38
rs782319177 Chr.19: 41444263-41444263 on Build GRCh38
rs369926537 Chr.19: 41444227-41444227 on Build GRCh38
rs3745285 Chr.19: 41444392-41444392 on Build GRCh38
rs17265865 Chr.19: 41444361-41444361 on Build GRCh38

View Full Product Details