Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168927125-168927398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCB11
Entrez Gene ID
8647
Gene Name
ATP binding cassette subfamily B member 11
Gene Aliases
ABC16, BRIC2, BSEP, PFIC-2, PFIC2, PGY4, SPGP
UniGene
Hs.658439
Gene Chromosome Location
Chr.2: 168915468-169031396 on Build GRCh38
Transcripts
NM_003742.2 XM_006712817.3 XM_011512077.2 XM_011512078.2 XM_011512081.2 XM_017005165.1 XM_017005166.1 XM_017005167.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7186762 c.3434A>G p.K1145R Chr.2: 168927340-168927340 on Build GRCh38
3314660 c.3613C>T p.P1205S Chr.2: 168927161-168927161 on Build GRCh38
6454613 c.3458G>A p.R1153H Chr.2: 168927316-168927316 on Build GRCh38
3407029 c.3573T>A p.A1191A Chr.2: 168927201-168927201 on Build GRCh38
1181293 c.3599T>G p.F1200C Chr.2: 168927175-168927175 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034148630 Chr.2: 168927280-168927280 on Build GRCh38
rs757040670 Chr.2: 168927369-168927369 on Build GRCh38
rs987819151 Chr.2: 168927349-168927349 on Build GRCh38
rs898922251 Chr.2: 168927169-168927169 on Build GRCh38
rs764069770 Chr.2: 168927257-168927257 on Build GRCh38
rs778415287 Chr.2: 168927314-168927314 on Build GRCh38
rs1521808 Chr.2: 168927218-168927218 on Build GRCh38
rs992165556 Chr.2: 168927307-168927307 on Build GRCh38
rs770257769 Chr.2: 168927301-168927301 on Build GRCh38
rs761682020 Chr.2: 168927353-168927353 on Build GRCh38
rs11568366 Chr.2: 168927339-168927339 on Build GRCh38
rs773929580 Chr.2: 168927320-168927320 on Build GRCh38
rs369313569 Chr.2: 168927325-168927325 on Build GRCh38
rs531704947 Chr.2: 168927345-168927345 on Build GRCh38
rs200709879 Chr.2: 168927250-168927250 on Build GRCh38
rs750541696 Chr.2: 168927261-168927261 on Build GRCh38
rs761903982 Chr.2: 168927312-168927312 on Build GRCh38
rs767761242 Chr.2: 168927372-168927372 on Build GRCh38
rs200735761 Chr.2: 168927348-168927348 on Build GRCh38
rs183621659 Chr.2: 168927262-168927262 on Build GRCh38
rs748967972 Chr.2: 168927352-168927352 on Build GRCh38
rs775198125 Chr.2: 168927341-168927341 on Build GRCh38
rs755163960 Chr.2: 168927271-168927271 on Build GRCh38
rs367984611 Chr.2: 168927147-168927147 on Build GRCh38
rs887349193 Chr.2: 168927228-168927228 on Build GRCh38
rs199762750 Chr.2: 168927266-168927266 on Build GRCh38
rs367730178 Chr.2: 168927313-168927313 on Build GRCh38
rs759204797 Chr.2: 168927232-168927232 on Build GRCh38
rs371084269 Chr.2: 168927148-168927148 on Build GRCh38
rs759292690 Chr.2: 168927335-168927335 on Build GRCh38
rs781443931 Chr.2: 168927273-168927273 on Build GRCh38
rs748862206 Chr.2: 168927316-168927316 on Build GRCh38
rs755647308 Chr.2: 168927283-168927283 on Build GRCh38
rs748499277 Chr.2: 168927285-168927285 on Build GRCh38
rs748610515 Chr.2: 168927206-168927206 on Build GRCh38
rs770770132 Chr.2: 168927225-168927225 on Build GRCh38
rs772623963 Chr.2: 168927217-168927217 on Build GRCh38
rs767218250 Chr.2: 168927247-168927247 on Build GRCh38
rs755107806 Chr.2: 168927255-168927255 on Build GRCh38
rs143484849 Chr.2: 168927226-168927226 on Build GRCh38
rs917920822 Chr.2: 168927311-168927311 on Build GRCh38
rs770299293 Chr.2: 168927207-168927207 on Build GRCh38
rs755181311 Chr.2: 168927370-168927370 on Build GRCh38
rs765282561 Chr.2: 168927356-168927356 on Build GRCh38
rs766453646 Chr.2: 168927361-168927361 on Build GRCh38
rs72549395 Chr.2: 168927317-168927317 on Build GRCh38
rs776566231 Chr.2: 168927167-168927167 on Build GRCh38
rs201481590 Chr.2: 168927268-168927268 on Build GRCh38

View Full Product Details