Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 27465301-27465570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF3C1
Entrez Gene ID
2975
Gene Name
general transcription factor IIIC subunit 1
Gene Aliases
TFIIIC, TFIIIC220, TFIIICalpha
UniGene
Hs.371718
Gene Chromosome Location
Chr.16: 27460613-27549930 on Build GRCh38
Transcripts
NM_001286242.1 NM_001520.3 XM_017023188.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4059641 c.5282A>C p.E1761A Chr.16: 27465333-27465333 on Build GRCh38
6867325 c.5188G>A p.E1730K Chr.16: 27465427-27465427 on Build GRCh38
6443485 c.5192T>G p.L1731R Chr.16: 27465423-27465423 on Build GRCh38
5885465 c.5178C>T p.V1726V Chr.16: 27465437-27465437 on Build GRCh38
4442007 c.5293C>T p.R1765W Chr.16: 27465322-27465322 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376298513 Chr.16: 27465327-27465327 on Build GRCh38
rs769269826 Chr.16: 27465448-27465448 on Build GRCh38
rs143331742 Chr.16: 27465447-27465447 on Build GRCh38
rs142565905 Chr.16: 27465407-27465407 on Build GRCh38
rs775811702 Chr.16: 27465336-27465336 on Build GRCh38
rs1047190781 Chr.16: 27465325-27465325 on Build GRCh38
rs374729096 Chr.16: 27465406-27465406 on Build GRCh38
rs764278754 Chr.16: 27465358-27465358 on Build GRCh38
rs750875265 Chr.16: 27465395-27465395 on Build GRCh38
rs764475050 Chr.16: 27465482-27465482 on Build GRCh38
rs1024351630 Chr.16: 27465399-27465399 on Build GRCh38
rs780871183 Chr.16: 27465505-27465505 on Build GRCh38
rs141586202 Chr.16: 27465491-27465491 on Build GRCh38
rs777097330 Chr.16: 27465527-27465527 on Build GRCh38
rs145495739 Chr.16: 27465350-27465350 on Build GRCh38
rs766780120 Chr.16: 27465500-27465500 on Build GRCh38
rs200068262 Chr.16: 27465334-27465334 on Build GRCh38
rs773956401 Chr.16: 27465477-27465477 on Build GRCh38
rs763321018 Chr.16: 27465499-27465499 on Build GRCh38
rs912256648 Chr.16: 27465526-27465526 on Build GRCh38
rs770703342 Chr.16: 27465539-27465539 on Build GRCh38
rs750506954 Chr.16: 27465385-27465385 on Build GRCh38
rs958663464 Chr.16: 27465387-27465387 on Build GRCh38
rs758841735 Chr.16: 27465400-27465400 on Build GRCh38
rs751213958 Chr.16: 27465322-27465322 on Build GRCh38
rs748092906 Chr.16: 27465321-27465321 on Build GRCh38
rs552379624 Chr.16: 27465360-27465360 on Build GRCh38
rs760978700 Chr.16: 27465481-27465481 on Build GRCh38
rs765203442 Chr.16: 27465496-27465496 on Build GRCh38
rs749257962 Chr.16: 27465538-27465538 on Build GRCh38
rs1019395168 Chr.16: 27465340-27465340 on Build GRCh38
rs377512725 Chr.16: 27465492-27465492 on Build GRCh38
rs753900389 Chr.16: 27465359-27465359 on Build GRCh38
rs1012914070 Chr.16: 27465394-27465394 on Build GRCh38
rs755652490 Chr.16: 27465519-27465519 on Build GRCh38
rs780687269 Chr.16: 27465425-27465425 on Build GRCh38
rs773239212 Chr.16: 27465459-27465459 on Build GRCh38
rs770781642 Chr.16: 27465468-27465468 on Build GRCh38
rs201984907 Chr.16: 27465516-27465516 on Build GRCh38
rs751806267 Chr.16: 27465502-27465502 on Build GRCh38
rs970781235 Chr.16: 27465535-27465535 on Build GRCh38
rs565913877 Chr.16: 27465373-27465373 on Build GRCh38
rs755677632 Chr.16: 27465505-27465507 on Build GRCh38
rs549734764 Chr.16: 27465324-27465324 on Build GRCh38
rs148341365 Chr.16: 27465463-27465463 on Build GRCh38
rs139970466 Chr.16: 27465328-27465328 on Build GRCh38

View Full Product Details