Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 7703338-7703611 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITIH2
Entrez Gene ID
3698
Gene Name
inter-alpha-trypsin inhibitor heavy chain 2
Gene Aliases
H2P, SHAP
UniGene
Hs.75285
Gene Chromosome Location
Chr.10: 7703273-7749520 on Build GRCh38
Transcripts
NM_002216.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6530765 c.38T>C p.L13P Chr.10: 7703472-7703472 on Build GRCh38
141283 c.58G>A p.E20K Chr.10: 7703492-7703492 on Build GRCh38
3440703 c.65C>T p.P22L Chr.10: 7703499-7703499 on Build GRCh38
3738730 c.84+3A>G p.? Chr.10: 7703521-7703521 on Build GRCh38
4557751 c.73G>A p.G25R Chr.10: 7703507-7703507 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187997136 Chr.10: 7703434-7703434 on Build GRCh38
rs926900984 Chr.10: 7703456-7703456 on Build GRCh38
rs747596906 Chr.10: 7703521-7703521 on Build GRCh38
rs777629047 Chr.10: 7703540-7703540 on Build GRCh38
rs764062742 Chr.10: 7703399-7703399 on Build GRCh38
rs754822796 Chr.10: 7703514-7703514 on Build GRCh38
rs2298319 Chr.10: 7703529-7703529 on Build GRCh38
rs767561816 Chr.10: 7703563-7703563 on Build GRCh38
rs550331397 Chr.10: 7703541-7703541 on Build GRCh38
rs375261970 Chr.10: 7703437-7703437 on Build GRCh38
rs367809007 Chr.10: 7703413-7703413 on Build GRCh38
rs753143542 Chr.10: 7703396-7703396 on Build GRCh38
rs776118079 Chr.10: 7703465-7703465 on Build GRCh38
rs890178410 Chr.10: 7703558-7703558 on Build GRCh38
rs549295916 Chr.10: 7703491-7703491 on Build GRCh38
rs777140767 Chr.10: 7703459-7703459 on Build GRCh38
rs141898195 Chr.10: 7703442-7703442 on Build GRCh38
rs201618342 Chr.10: 7703554-7703554 on Build GRCh38
rs35587469 Chr.10: 7703582-7703583 on Build GRCh38
rs34117267 Chr.10: 7703448-7703448 on Build GRCh38
rs200009045 Chr.10: 7703499-7703499 on Build GRCh38
rs866857197 Chr.10: 7703408-7703408 on Build GRCh38
rs200957680 Chr.10: 7703418-7703418 on Build GRCh38
rs770271447 Chr.10: 7703461-7703461 on Build GRCh38
rs150632269 Chr.10: 7703449-7703449 on Build GRCh38
rs771794108 Chr.10: 7703453-7703453 on Build GRCh38
rs372204187 Chr.10: 7703546-7703546 on Build GRCh38
rs34028333 Chr.10: 7703571-7703572 on Build GRCh38
rs760135551 Chr.10: 7703389-7703389 on Build GRCh38
rs763341525 Chr.10: 7703474-7703474 on Build GRCh38
rs778500218 Chr.10: 7703515-7703515 on Build GRCh38
rs538091209 Chr.10: 7703500-7703500 on Build GRCh38
rs774232752 Chr.10: 7703481-7703481 on Build GRCh38
rs774831508 Chr.10: 7703556-7703556 on Build GRCh38
rs267602582 Chr.10: 7703492-7703492 on Build GRCh38
rs761937267 Chr.10: 7703557-7703557 on Build GRCh38
rs201922110 Chr.10: 7703395-7703395 on Build GRCh38
rs143303387 Chr.10: 7703489-7703489 on Build GRCh38
rs755951323 Chr.10: 7703497-7703497 on Build GRCh38
rs780975495 Chr.10: 7703432-7703432 on Build GRCh38
rs1007415756 Chr.10: 7703586-7703586 on Build GRCh38
rs183350466 Chr.10: 7703397-7703397 on Build GRCh38
rs765774340 Chr.10: 7703391-7703391 on Build GRCh38
rs764252763 Chr.10: 7703475-7703475 on Build GRCh38
rs760047689 Chr.10: 7703460-7703460 on Build GRCh38
rs942263876 Chr.10: 7703483-7703483 on Build GRCh38
rs1037408132 Chr.10: 7703509-7703509 on Build GRCh38

View Full Product Details