Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30927233-30927460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXL19
Entrez Gene ID
54620
Gene Name
F-box and leucine rich repeat protein 19
Gene Aliases
CXXC11, Fbl19, JHDM1C
UniGene
Hs.152149
Gene Chromosome Location
Chr.16: 30923055-30948783 on Build GRCh38
Transcripts
NM_001099784.2 NM_001282351.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5850762 c.341G>A p.C114Y Chr.16: 30927411-30927411 on Build GRCh38
5850761 c.298G>A p.V100M Chr.16: 30927368-30927368 on Build GRCh38
6248371 c.355G>T p.E119* Chr.16: 30927425-30927425 on Build GRCh38
6324911 c.313G>A p.E105K Chr.16: 30927383-30927383 on Build GRCh38
5384761 c.256G>A p.A86T Chr.16: 30927326-30927326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568469822 Chr.16: 30927264-30927264 on Build GRCh38
rs910358942 Chr.16: 30927367-30927367 on Build GRCh38
rs761238581 Chr.16: 30927292-30927292 on Build GRCh38
rs943141682 Chr.16: 30927399-30927399 on Build GRCh38
rs878987159 Chr.16: 30927280-30927280 on Build GRCh38
rs369393189 Chr.16: 30927331-30927331 on Build GRCh38
rs35973407 Chr.16: 30927262-30927263 on Build GRCh38
rs752093184 Chr.16: 30927276-30927276 on Build GRCh38
rs758592580 Chr.16: 30927431-30927431 on Build GRCh38
rs762322417 Chr.16: 30927272-30927272 on Build GRCh38
rs529354031 Chr.16: 30927304-30927304 on Build GRCh38
rs756524918 Chr.16: 30927318-30927333 on Build GRCh38
rs373327143 Chr.16: 30927439-30927439 on Build GRCh38
rs983271232 Chr.16: 30927436-30927436 on Build GRCh38
rs201684326 Chr.16: 30927337-30927337 on Build GRCh38
rs750676545 Chr.16: 30927424-30927424 on Build GRCh38
rs765511827 Chr.16: 30927373-30927373 on Build GRCh38
rs751070040 Chr.16: 30927277-30927277 on Build GRCh38
rs376844315 Chr.16: 30927441-30927441 on Build GRCh38

View Full Product Details