Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133267275-133267502 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM8
Entrez Gene ID
101
Gene Name
ADAM metallopeptidase domain 8
Gene Aliases
CD156, CD156a, MS2
UniGene
Hs.501574
Gene Chromosome Location
Chr.10: 133262416-133276903 on Build GRCh38
Transcripts
NM_001109.4 NM_001164489.1 NM_001164490.1 XM_011539117.2 XM_017015465.1

SNP Detail

SNP ID Chromosome Location
rs971941345 Chr.10: 133267449-133267449 on Build GRCh38
rs376925289 Chr.10: 133267368-133267368 on Build GRCh38
rs766840898 Chr.10: 133267379-133267379 on Build GRCh38
rs773539219 Chr.10: 133267371-133267371 on Build GRCh38
rs532126257 Chr.10: 133267381-133267381 on Build GRCh38
rs777358233 Chr.10: 133267325-133267325 on Build GRCh38
rs752275497 Chr.10: 133267404-133267404 on Build GRCh38
rs574716445 Chr.10: 133267340-133267340 on Build GRCh38
rs377741089 Chr.10: 133267306-133267306 on Build GRCh38
rs531327978 Chr.10: 133267466-133267466 on Build GRCh38
rs368744832 Chr.10: 133267382-133267382 on Build GRCh38
rs756095777 Chr.10: 133267462-133267462 on Build GRCh38
rs150169740 Chr.10: 133267412-133267412 on Build GRCh38
rs140865131 Chr.10: 133267373-133267373 on Build GRCh38
rs113224991 Chr.10: 133267456-133267456 on Build GRCh38
rs866854575 Chr.10: 133267406-133267406 on Build GRCh38
rs370892454 Chr.10: 133267314-133267314 on Build GRCh38
rs544391181 Chr.10: 133267323-133267323 on Build GRCh38
rs767096516 Chr.10: 133267374-133267375 on Build GRCh38
rs1033597620 Chr.10: 133267399-133267399 on Build GRCh38
rs777479740 Chr.10: 133267397-133267397 on Build GRCh38
rs1023925227 Chr.10: 133267420-133267420 on Build GRCh38
rs952071592 Chr.10: 133267459-133267459 on Build GRCh38
rs371362381 Chr.10: 133267434-133267434 on Build GRCh38
rs550728560 Chr.10: 133267471-133267471 on Build GRCh38
rs761746442 Chr.10: 133267361-133267361 on Build GRCh38
rs558967916 Chr.10: 133267301-133267301 on Build GRCh38
rs942781548 Chr.10: 133267328-133267328 on Build GRCh38
rs772259908 Chr.10: 133267467-133267467 on Build GRCh38
rs759320307 Chr.10: 133267408-133267408 on Build GRCh38
rs903447485 Chr.10: 133267395-133267395 on Build GRCh38
rs745801799 Chr.10: 133267339-133267339 on Build GRCh38
rs751199492 Chr.10: 133267435-133267435 on Build GRCh38
rs5789158 Chr.10: 133267476-133267477 on Build GRCh38
rs761255645 Chr.10: 133267369-133267369 on Build GRCh38
rs757110020 Chr.10: 133267436-133267436 on Build GRCh38
rs1033007104 Chr.10: 133267385-133267385 on Build GRCh38
rs758084453 Chr.10: 133267324-133267324 on Build GRCh38
rs79495431 Chr.10: 133267369-133267369 on Build GRCh38
rs774498849 Chr.10: 133267342-133267342 on Build GRCh38
rs746844175 Chr.10: 133267330-133267330 on Build GRCh38
rs368644961 Chr.10: 133267331-133267331 on Build GRCh38
rs375332080 Chr.10: 133267437-133267437 on Build GRCh38
rs545492149 Chr.10: 133267413-133267413 on Build GRCh38
rs2860375 Chr.10: 133267322-133267322 on Build GRCh38
rs971062175 Chr.10: 133267421-133267421 on Build GRCh38
rs779813556 Chr.10: 133267463-133267463 on Build GRCh38
rs765159308 Chr.10: 133267447-133267447 on Build GRCh38
rs1012655949 Chr.10: 133267414-133267414 on Build GRCh38

View Full Product Details