Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130341822-130342047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC115
Entrez Gene ID
84317
Gene Name
coiled-coil domain containing 115
Gene Aliases
CDG2O, ccp1
UniGene
Hs.104203
Gene Chromosome Location
Chr.2: 130337933-130342681 on Build GRCh38
Transcripts
NM_001321118.1 NM_001321119.1 NM_032357.3 NR_104471.1 NR_135548.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4084817 c.97G>T p.G33C Chr.2: 130341944-130341944 on Build GRCh38
279613 c.104T>C p.V35A Chr.2: 130342022-130342022 on Build GRCh38
6621344 c.96C>T p.N32N Chr.2: 130342030-130342030 on Build GRCh38
2156256 c.112-3C>A p.? Chr.2: 130341947-130341947 on Build GRCh38
7304503 c.160T>C p.Y54H Chr.2: 130341881-130341881 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1035273120 Chr.2: 130341997-130341998 on Build GRCh38
rs753725482 Chr.2: 130341888-130341888 on Build GRCh38
rs745621758 Chr.2: 130341897-130341897 on Build GRCh38
rs376279552 Chr.2: 130341982-130341982 on Build GRCh38
rs372936718 Chr.2: 130341873-130341873 on Build GRCh38
rs376570015 Chr.2: 130341968-130341968 on Build GRCh38
rs865959683 Chr.2: 130341874-130341874 on Build GRCh38
rs937974515 Chr.2: 130341961-130341961 on Build GRCh38
rs1043767273 Chr.2: 130341903-130341903 on Build GRCh38
rs763766228 Chr.2: 130342010-130342010 on Build GRCh38
rs765309029 Chr.2: 130341946-130341946 on Build GRCh38
rs775073648 Chr.2: 130341929-130341929 on Build GRCh38
rs199767164 Chr.2: 130341984-130341984 on Build GRCh38
rs985824030 Chr.2: 130341851-130341851 on Build GRCh38
rs776120445 Chr.2: 130342011-130342011 on Build GRCh38
rs1003677034 Chr.2: 130341995-130341995 on Build GRCh38
rs776711440 Chr.2: 130341917-130341917 on Build GRCh38
rs753340310 Chr.2: 130341938-130341938 on Build GRCh38
rs771291770 Chr.2: 130341996-130341996 on Build GRCh38
rs750387342 Chr.2: 130342023-130342023 on Build GRCh38
rs577686738 Chr.2: 130341989-130341989 on Build GRCh38
rs766713863 Chr.2: 130341922-130341922 on Build GRCh38
rs761461065 Chr.2: 130342018-130342018 on Build GRCh38
rs756519662 Chr.2: 130341975-130341975 on Build GRCh38
rs910788027 Chr.2: 130341863-130341863 on Build GRCh38
rs769842324 Chr.2: 130341986-130341986 on Build GRCh38
rs774721477 Chr.2: 130342001-130342001 on Build GRCh38
rs779925212 Chr.2: 130341956-130341956 on Build GRCh38
rs375016127 Chr.2: 130341845-130341845 on Build GRCh38
rs750366846 Chr.2: 130341951-130341951 on Build GRCh38
rs370873767 Chr.2: 130341974-130341974 on Build GRCh38
rs780475360 Chr.2: 130341908-130341908 on Build GRCh38
rs758328738 Chr.2: 130341954-130341954 on Build GRCh38
rs894366722 Chr.2: 130341970-130341970 on Build GRCh38
rs556226856 Chr.2: 130341842-130341842 on Build GRCh38
rs760268834 Chr.2: 130342008-130342008 on Build GRCh38
rs151154806 Chr.2: 130341920-130341920 on Build GRCh38
rs771965486 Chr.2: 130341905-130341905 on Build GRCh38
rs763525735 Chr.2: 130341930-130341930 on Build GRCh38
rs777335139 Chr.2: 130341885-130341885 on Build GRCh38
rs778877117 Chr.2: 130341891-130341891 on Build GRCh38
rs750733183 Chr.2: 130341866-130341866 on Build GRCh38
rs914098043 Chr.2: 130341889-130341889 on Build GRCh38
rs748620732 Chr.2: 130341919-130341919 on Build GRCh38
rs576449724 Chr.2: 130341910-130341910 on Build GRCh38
rs764828737 Chr.2: 130342019-130342019 on Build GRCh38
rs768620514 Chr.2: 130341912-130341912 on Build GRCh38
rs201329717 Chr.2: 130341948-130341948 on Build GRCh38
rs755635296 Chr.2: 130341877-130341877 on Build GRCh38
rs757177047 Chr.2: 130341943-130341943 on Build GRCh38
rs749789810 Chr.2: 130341990-130341990 on Build GRCh38

View Full Product Details