Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 50446379-50446638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIGNL1
Entrez Gene ID
63979
Gene Name
fidgetin like 1
Gene Aliases
-
UniGene
Hs.137516
Gene Chromosome Location
Chr.7: 50437327-50450390 on Build GRCh38
Transcripts
NM_001042762.2 NM_001287492.1 NM_001287493.1 NM_001287494.1 NM_001287495.1 NM_001287496.1 NM_022116.4 XM_011515470.2 XM_017012501.1 XM_017012502.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6028953 c.830C>T p.A277V Chr.7: 50446458-50446458 on Build GRCh38
1090597 c.661T>C p.F221L Chr.7: 50446627-50446627 on Build GRCh38
6480772 c.781T>C p.Y261H Chr.7: 50446507-50446507 on Build GRCh38
6861435 c.692C>G p.S231C Chr.7: 50446596-50446596 on Build GRCh38
1090596 c.689G>T p.S230I Chr.7: 50446599-50446599 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767305212 Chr.7: 50446486-50446486 on Build GRCh38
rs79851593 Chr.7: 50446506-50446506 on Build GRCh38
rs775331601 Chr.7: 50446487-50446487 on Build GRCh38
rs113757480 Chr.7: 50446476-50446476 on Build GRCh38
rs760598195 Chr.7: 50446598-50446598 on Build GRCh38
rs556446618 Chr.7: 50446596-50446596 on Build GRCh38
rs374660466 Chr.7: 50446577-50446577 on Build GRCh38
rs781736431 Chr.7: 50446561-50446561 on Build GRCh38
rs114977235 Chr.7: 50446471-50446471 on Build GRCh38
rs769234078 Chr.7: 50446470-50446470 on Build GRCh38
rs762669958 Chr.7: 50446419-50446419 on Build GRCh38
rs144012928 Chr.7: 50446438-50446438 on Build GRCh38
rs750656597 Chr.7: 50446611-50446611 on Build GRCh38
rs756179794 Chr.7: 50446440-50446440 on Build GRCh38
rs376193207 Chr.7: 50446489-50446489 on Build GRCh38
rs772427310 Chr.7: 50446467-50446467 on Build GRCh38
rs139245614 Chr.7: 50446570-50446570 on Build GRCh38
rs146689210 Chr.7: 50446459-50446459 on Build GRCh38
rs759273253 Chr.7: 50446480-50446480 on Build GRCh38
rs144297981 Chr.7: 50446599-50446599 on Build GRCh38
rs370131380 Chr.7: 50446578-50446578 on Build GRCh38
rs140225959 Chr.7: 50446469-50446469 on Build GRCh38
rs765462822 Chr.7: 50446511-50446511 on Build GRCh38
rs143316695 Chr.7: 50446491-50446491 on Build GRCh38
rs761908505 Chr.7: 50446601-50446601 on Build GRCh38
rs375386461 Chr.7: 50446434-50446434 on Build GRCh38
rs746240766 Chr.7: 50446465-50446465 on Build GRCh38
rs184297998 Chr.7: 50446539-50446539 on Build GRCh38
rs754110258 Chr.7: 50446453-50446453 on Build GRCh38
rs752268516 Chr.7: 50446543-50446544 on Build GRCh38
rs865847820 Chr.7: 50446521-50446522 on Build GRCh38
rs765266283 Chr.7: 50446609-50446609 on Build GRCh38
rs371326486 Chr.7: 50446479-50446479 on Build GRCh38
rs549218107 Chr.7: 50446420-50446420 on Build GRCh38
rs750613807 Chr.7: 50446517-50446517 on Build GRCh38
rs759307879 Chr.7: 50446432-50446432 on Build GRCh38
rs751393424 Chr.7: 50446424-50446424 on Build GRCh38
rs369555128 Chr.7: 50446542-50446542 on Build GRCh38
rs753913264 Chr.7: 50446503-50446503 on Build GRCh38
rs893109840 Chr.7: 50446515-50446515 on Build GRCh38
rs762758199 Chr.7: 50446473-50446473 on Build GRCh38
rs149596053 Chr.7: 50446581-50446581 on Build GRCh38
rs757460815 Chr.7: 50446455-50446455 on Build GRCh38
rs113845281 Chr.7: 50446521-50446521 on Build GRCh38
rs777801333 Chr.7: 50446451-50446451 on Build GRCh38
rs1020511186 Chr.7: 50446522-50446522 on Build GRCh38
rs889657235 Chr.7: 50446472-50446472 on Build GRCh38

View Full Product Details