Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 119570494-119570753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENPP2
Entrez Gene ID
5168
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 2
Gene Aliases
ATX, ATX-X, AUTOTAXIN, LysoPLD, NPP2, PD-IALPHA, PDNP2
UniGene
Hs.190977
Gene Chromosome Location
Chr.8: 119557077-119673404 on Build GRCh38
Transcripts
NM_001040092.2 NM_001130863.2 NM_006209.4 NR_045555.1 XM_006716584.1 XM_006716585.1 XM_006716587.1 XM_017013571.1 XM_017013572.1 XM_017013573.1 XM_017013574.1 XM_017013575.1

SNP Detail

SNP ID Chromosome Location
rs746404728 Chr.8: 119570663-119570663 on Build GRCh38
rs760483974 Chr.8: 119570720-119570720 on Build GRCh38
rs762531230 Chr.8: 119570691-119570692 on Build GRCh38
rs16892786 Chr.8: 119570631-119570631 on Build GRCh38
rs186915683 Chr.8: 119570635-119570635 on Build GRCh38
rs770672175 Chr.8: 119570727-119570727 on Build GRCh38
rs1019887113 Chr.8: 119570603-119570603 on Build GRCh38
rs771716771 Chr.8: 119570700-119570700 on Build GRCh38
rs899976005 Chr.8: 119570604-119570604 on Build GRCh38
rs113629030 Chr.8: 119570633-119570633 on Build GRCh38
rs778639049 Chr.8: 119570578-119570578 on Build GRCh38
rs901044834 Chr.8: 119570673-119570673 on Build GRCh38
rs1008880656 Chr.8: 119570595-119570595 on Build GRCh38
rs1039641690 Chr.8: 119570659-119570659 on Build GRCh38
rs778346317 Chr.8: 119570677-119570677 on Build GRCh38
rs549481860 Chr.8: 119570591-119570591 on Build GRCh38
rs371783576 Chr.8: 119570689-119570689 on Build GRCh38
rs770315463 Chr.8: 119570674-119570674 on Build GRCh38
rs781773243 Chr.8: 119570657-119570657 on Build GRCh38
rs758079918 Chr.8: 119570687-119570687 on Build GRCh38
rs1004342361 Chr.8: 119570701-119570701 on Build GRCh38
rs1007944795 Chr.8: 119570655-119570655 on Build GRCh38
rs531649331 Chr.8: 119570587-119570587 on Build GRCh38
rs772793217 Chr.8: 119570716-119570716 on Build GRCh38
rs76900353 Chr.8: 119570558-119570558 on Build GRCh38

View Full Product Details