Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 185744153-185744400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSIP2
Entrez Gene ID
401024
Gene Name
fibrous sheath interacting protein 2
Gene Aliases
-
UniGene
Hs.98025
Gene Chromosome Location
Chr.2: 185738895-185833289 on Build GRCh38
Transcripts
NM_173651.3

SNP Detail

SNP ID Chromosome Location
rs1038936870 Chr.2: 185744286-185744286 on Build GRCh38
rs977854447 Chr.2: 185744247-185744247 on Build GRCh38
rs901775124 Chr.2: 185744296-185744296 on Build GRCh38
rs887645389 Chr.2: 185744240-185744240 on Build GRCh38
rs943185388 Chr.2: 185744249-185744249 on Build GRCh38
rs912226128 Chr.2: 185744226-185744233 on Build GRCh38
rs111319757 Chr.2: 185744220-185744220 on Build GRCh38
rs769794876 Chr.2: 185744274-185744274 on Build GRCh38
rs562668559 Chr.2: 185744325-185744325 on Build GRCh38
rs965450536 Chr.2: 185744227-185744227 on Build GRCh38
rs7565249 Chr.2: 185744351-185744351 on Build GRCh38
rs985050446 Chr.2: 185744210-185744212 on Build GRCh38
rs754614966 Chr.2: 185744313-185744313 on Build GRCh38
rs1053631208 Chr.2: 185744354-185744354 on Build GRCh38

View Full Product Details