Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 175441885-175442140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD1
Entrez Gene ID
1812
Gene Name
dopamine receptor D1
Gene Aliases
DADR, DRD1A
UniGene
Hs.2624
Gene Chromosome Location
Chr.5: 175440672-175444160 on Build GRCh38
Transcripts
NM_000794.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3614247 c.1126G>A p.E376K Chr.5: 175441974-175441974 on Build GRCh38
4457668 c.1054C>T p.P352S Chr.5: 175442046-175442046 on Build GRCh38
5647424 c.1177C>T p.H393Y Chr.5: 175441923-175441923 on Build GRCh38
3614248 c.1107G>A p.A369A Chr.5: 175441993-175441993 on Build GRCh38
5699332 c.1079C>A p.T360K Chr.5: 175442021-175442021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771065562 Chr.5: 175442058-175442058 on Build GRCh38
rs528645013 Chr.5: 175442013-175442013 on Build GRCh38
rs767938936 Chr.5: 175441917-175441917 on Build GRCh38
rs774488532 Chr.5: 175442071-175442071 on Build GRCh38
rs141925440 Chr.5: 175442116-175442116 on Build GRCh38
rs760024652 Chr.5: 175442002-175442002 on Build GRCh38
rs745758211 Chr.5: 175441965-175441965 on Build GRCh38
rs753165553 Chr.5: 175442019-175442020 on Build GRCh38
rs532843195 Chr.5: 175442088-175442088 on Build GRCh38
rs763112127 Chr.5: 175441995-175441995 on Build GRCh38
rs755465400 Chr.5: 175441912-175441912 on Build GRCh38
rs776180680 Chr.5: 175442113-175442113 on Build GRCh38
rs761172175 Chr.5: 175442015-175442015 on Build GRCh38
rs763809199 Chr.5: 175442020-175442020 on Build GRCh38
rs753491559 Chr.5: 175442021-175442021 on Build GRCh38
rs373118849 Chr.5: 175441996-175441996 on Build GRCh38
rs777626977 Chr.5: 175441946-175441946 on Build GRCh38
rs768208899 Chr.5: 175442011-175442011 on Build GRCh38
rs374239045 Chr.5: 175441993-175441993 on Build GRCh38
rs746864254 Chr.5: 175441986-175441986 on Build GRCh38
rs753120440 Chr.5: 175441922-175441922 on Build GRCh38
rs201076076 Chr.5: 175441994-175441994 on Build GRCh38
rs748726799 Chr.5: 175441947-175441947 on Build GRCh38
rs756834255 Chr.5: 175441951-175441951 on Build GRCh38
rs768398169 Chr.5: 175442054-175442054 on Build GRCh38
rs144813919 Chr.5: 175442042-175442042 on Build GRCh38
rs6272 Chr.5: 175442056-175442056 on Build GRCh38
rs370098478 Chr.5: 175442053-175442053 on Build GRCh38
rs765528098 Chr.5: 175441913-175441920 on Build GRCh38
rs780868574 Chr.5: 175442055-175442055 on Build GRCh38
rs775260576 Chr.5: 175441979-175441979 on Build GRCh38
rs759873198 Chr.5: 175441911-175441911 on Build GRCh38
rs756872143 Chr.5: 175442038-175442038 on Build GRCh38
rs772685461 Chr.5: 175442074-175442074 on Build GRCh38
rs771907312 Chr.5: 175441967-175441967 on Build GRCh38
rs778418674 Chr.5: 175442039-175442039 on Build GRCh38
rs749967573 Chr.5: 175442041-175442041 on Build GRCh38
rs778319909 Chr.5: 175441964-175441964 on Build GRCh38
rs755827909 Chr.5: 175441938-175441938 on Build GRCh38

View Full Product Details