Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36322632-36322857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EVA1B
Entrez Gene ID
55194
Gene Name
eva-1 homolog B (C. elegans)
Gene Aliases
C1orf78, FAM176B
UniGene
Hs.87016
Gene Chromosome Location
Chr.1: 36322030-36324154 on Build GRCh38
Transcripts
NM_001304762.1 NM_018166.2 NR_130899.1 XM_017001627.1 XM_017002340.1 XM_017002341.1 XM_017002344.1 XM_017002345.1

Gene Symbol
SH3D21
Entrez Gene ID
79729
Gene Name
SH3 domain containing 21
Gene Aliases
C1orf113
UniGene
Hs.524496
Gene Chromosome Location
Chr.1: 36306348-36329221 on Build GRCh38
Transcripts
NM_001304762.1 NM_018166.2 NR_130899.1 XM_017001627.1 XM_017002340.1 XM_017002341.1 XM_017002344.1 XM_017002345.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1342303 c.76G>A p.E26K Chr.1: 36322717-36322717 on Build GRCh38
6042213 c.105C>T p.G35G Chr.1: 36322688-36322688 on Build GRCh38
5550138 c.161C>T p.A54V Chr.1: 36322632-36322632 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs113936116 Chr.1: 36322827-36322827 on Build GRCh38
rs888540820 Chr.1: 36322675-36322675 on Build GRCh38
rs919202620 Chr.1: 36322838-36322838 on Build GRCh38
rs7536394 Chr.1: 36322755-36322755 on Build GRCh38
rs1034370838 Chr.1: 36322725-36322725 on Build GRCh38
rs768316044 Chr.1: 36322688-36322688 on Build GRCh38
rs768703283 Chr.1: 36322752-36322752 on Build GRCh38
rs766889776 Chr.1: 36322775-36322775 on Build GRCh38
rs985813761 Chr.1: 36322798-36322798 on Build GRCh38
rs929154659 Chr.1: 36322733-36322733 on Build GRCh38
rs758069931 Chr.1: 36322748-36322748 on Build GRCh38
rs896463343 Chr.1: 36322765-36322765 on Build GRCh38
rs777572883 Chr.1: 36322745-36322745 on Build GRCh38
rs751232112 Chr.1: 36322777-36322777 on Build GRCh38
rs536998405 Chr.1: 36322751-36322751 on Build GRCh38
rs887621770 Chr.1: 36322743-36322743 on Build GRCh38
rs1000965000 Chr.1: 36322797-36322797 on Build GRCh38
rs746798396 Chr.1: 36322800-36322800 on Build GRCh38
rs1002037077 Chr.1: 36322744-36322744 on Build GRCh38
rs576634477 Chr.1: 36322819-36322819 on Build GRCh38
rs756037850 Chr.1: 36322701-36322701 on Build GRCh38
rs773606790 Chr.1: 36322770-36322770 on Build GRCh38
rs780801449 Chr.1: 36322654-36322654 on Build GRCh38
rs1026452051 Chr.1: 36322687-36322687 on Build GRCh38
rs1049403603 Chr.1: 36322736-36322736 on Build GRCh38
rs965682593 Chr.1: 36322836-36322836 on Build GRCh38
rs763397436 Chr.1: 36322771-36322771 on Build GRCh38
rs910288595 Chr.1: 36322816-36322816 on Build GRCh38
rs954209080 Chr.1: 36322810-36322810 on Build GRCh38
rs749050710 Chr.1: 36322746-36322746 on Build GRCh38
rs754506123 Chr.1: 36322652-36322652 on Build GRCh38
rs770270546 Chr.1: 36322764-36322764 on Build GRCh38
rs752170638 Chr.1: 36322700-36322700 on Build GRCh38

View Full Product Details