Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113424910-113425183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf43
Entrez Gene ID
257169
Gene Name
chromosome 9 open reading frame 43
Gene Aliases
-
UniGene
Hs.632691
Gene Chromosome Location
Chr.9: 113410644-113429684 on Build GRCh38
Transcripts
NM_001278629.1 NM_001278630.1 NM_152786.2 XM_005251885.3 XM_011518477.1 XM_011518478.1 XM_011518479.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6237148 c.808-10C>T p.? Chr.9: 113425009-113425009 on Build GRCh38
3903231 c.832C>T p.R278C Chr.9: 113425043-113425043 on Build GRCh38
1314384 c.833G>T p.R278L Chr.9: 113425044-113425044 on Build GRCh38
7076306 c.808-2A>G p.? Chr.9: 113425017-113425017 on Build GRCh38
3320208 c.820C>T p.R274* Chr.9: 113425031-113425031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963575833 Chr.9: 113425027-113425027 on Build GRCh38
rs765201166 Chr.9: 113425009-113425009 on Build GRCh38
rs571864386 Chr.9: 113425050-113425050 on Build GRCh38
rs186682268 Chr.9: 113425139-113425139 on Build GRCh38
rs767647074 Chr.9: 113425114-113425129 on Build GRCh38
rs774951478 Chr.9: 113425059-113425059 on Build GRCh38
rs763891949 Chr.9: 113425001-113425001 on Build GRCh38
rs922620953 Chr.9: 113424945-113424945 on Build GRCh38
rs775742221 Chr.9: 113424990-113424990 on Build GRCh38
rs967864131 Chr.9: 113425080-113425080 on Build GRCh38
rs111748634 Chr.9: 113425022-113425022 on Build GRCh38
rs749909970 Chr.9: 113425012-113425012 on Build GRCh38
rs746409820 Chr.9: 113425061-113425061 on Build GRCh38
rs1011843365 Chr.9: 113424944-113424944 on Build GRCh38
rs772124466 Chr.9: 113425070-113425070 on Build GRCh38
rs376559063 Chr.9: 113424973-113424973 on Build GRCh38
rs561322739 Chr.9: 113425146-113425146 on Build GRCh38
rs200902357 Chr.9: 113425043-113425043 on Build GRCh38
rs761648943 Chr.9: 113425128-113425128 on Build GRCh38
rs759384919 Chr.9: 113425051-113425051 on Build GRCh38
rs775921675 Chr.9: 113424939-113424939 on Build GRCh38
rs774710537 Chr.9: 113425069-113425069 on Build GRCh38
rs201215268 Chr.9: 113425031-113425031 on Build GRCh38
rs762184190 Chr.9: 113425080-113425080 on Build GRCh38
rs776457568 Chr.9: 113425003-113425003 on Build GRCh38
rs746600411 Chr.9: 113425032-113425032 on Build GRCh38
rs749735235 Chr.9: 113425044-113425044 on Build GRCh38
rs764294069 Chr.9: 113425101-113425101 on Build GRCh38
rs759729078 Chr.9: 113424977-113424977 on Build GRCh38
rs765165667 Chr.9: 113425057-113425057 on Build GRCh38
rs953629805 Chr.9: 113425054-113425054 on Build GRCh38
rs775446349 Chr.9: 113425074-113425074 on Build GRCh38
rs760941483 Chr.9: 113425075-113425075 on Build GRCh38
rs28553410 Chr.9: 113425147-113425147 on Build GRCh38
rs527987421 Chr.9: 113424937-113424937 on Build GRCh38
rs771532354 Chr.9: 113425057-113425057 on Build GRCh38
rs760813516 Chr.9: 113424995-113424995 on Build GRCh38
rs753638688 Chr.9: 113425002-113425002 on Build GRCh38
rs1046075666 Chr.9: 113424955-113424955 on Build GRCh38
rs753395261 Chr.9: 113425145-113425145 on Build GRCh38
rs776382577 Chr.9: 113425107-113425107 on Build GRCh38
rs757185709 Chr.9: 113425003-113425003 on Build GRCh38
rs757460294 Chr.9: 113425064-113425064 on Build GRCh38
rs151254029 Chr.9: 113425037-113425037 on Build GRCh38
rs370303958 Chr.9: 113424975-113424975 on Build GRCh38
rs767730068 Chr.9: 113424985-113424985 on Build GRCh38
rs1038488184 Chr.9: 113425124-113425124 on Build GRCh38

View Full Product Details