Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 31121752-31122005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMD
Entrez Gene ID
1756
Gene Name
dystrophin
Gene Aliases
BMD, CMD3B, DXS142, DXS164, DXS206, DXS230, DXS239, DXS268, DXS269, DXS270, DXS272, MRX85
UniGene
Hs.495912
Gene Chromosome Location
Chr.X: 31119219-33339609 on Build GRCh38
Transcripts
NM_000109.3 NM_004006.2 NM_004009.3 NM_004010.3 NM_004011.3 NM_004012.3 NM_004013.2 NM_004014.2 NM_004015.2 NM_004016.2 NM_004017.2 NM_004018.2 NM_004020.3 NM_004021.2 NM_004022.2 NM_004023.2 XM_006724468.2 XM_006724469.3 XM_006724470.3 XM_006724473.2 XM_006724474.3 XM_006724475.2 XM_011545467.1 XM_017029328.1 XM_017029331.1

SNP Detail

SNP ID Chromosome Location
rs889509708 Chr.X: 31121846-31121846 on Build GRCh38
rs779159590 Chr.X: 31121974-31121977 on Build GRCh38
rs371730838 Chr.X: 31121880-31121880 on Build GRCh38
rs770298921 Chr.X: 31121830-31121830 on Build GRCh38
rs767488684 Chr.X: 31121868-31121868 on Build GRCh38
rs781341977 Chr.X: 31121952-31121955 on Build GRCh38
rs75075462 Chr.X: 31121787-31121787 on Build GRCh38
rs752332058 Chr.X: 31121884-31121896 on Build GRCh38
rs747862290 Chr.X: 31121810-31121810 on Build GRCh38
rs752529932 Chr.X: 31121871-31121871 on Build GRCh38
rs772389266 Chr.X: 31121973-31121973 on Build GRCh38
rs772681327 Chr.X: 31121811-31121811 on Build GRCh38
rs777504092 Chr.X: 31121907-31121907 on Build GRCh38
rs748558169 Chr.X: 31121902-31121902 on Build GRCh38
rs774279444 Chr.X: 31121841-31121841 on Build GRCh38
rs755056874 Chr.X: 31121973-31121974 on Build GRCh38
rs404401 Chr.X: 31121788-31121788 on Build GRCh38
rs772241320 Chr.X: 31121954-31121954 on Build GRCh38
rs757261855 Chr.X: 31121944-31121947 on Build GRCh38
rs377175004 Chr.X: 31121821-31121821 on Build GRCh38
rs772151654 Chr.X: 31121970-31121970 on Build GRCh38
rs759517642 Chr.X: 31121857-31121857 on Build GRCh38
rs773994505 Chr.X: 31121964-31121964 on Build GRCh38
rs149405184 Chr.X: 31121904-31121904 on Build GRCh38
rs372284841 Chr.X: 31121895-31121895 on Build GRCh38
rs755470544 Chr.X: 31121888-31121888 on Build GRCh38
rs1017766859 Chr.X: 31121903-31121903 on Build GRCh38
rs748870087 Chr.X: 31121910-31121910 on Build GRCh38
rs372759340 Chr.X: 31121783-31121783 on Build GRCh38
rs759027519 Chr.X: 31121788-31121791 on Build GRCh38
rs780081481 Chr.X: 31121882-31121882 on Build GRCh38
rs770516154 Chr.X: 31121917-31121917 on Build GRCh38
rs759607747 Chr.X: 31121969-31121969 on Build GRCh38
rs765504838 Chr.X: 31121878-31121878 on Build GRCh38
rs775505897 Chr.X: 31121980-31121980 on Build GRCh38
rs764000962 Chr.X: 31121940-31121943 on Build GRCh38
rs752098195 Chr.X: 31121887-31121887 on Build GRCh38
rs753936230 Chr.X: 31121965-31121966 on Build GRCh38
rs751483072 Chr.X: 31121940-31121941 on Build GRCh38
rs764921358 Chr.X: 31121861-31121866 on Build GRCh38
rs781502432 Chr.X: 31121799-31121799 on Build GRCh38
rs762767369 Chr.X: 31121910-31121922 on Build GRCh38
rs16989352 Chr.X: 31121881-31121881 on Build GRCh38
rs369622987 Chr.X: 31121824-31121824 on Build GRCh38
rs760332250 Chr.X: 31121874-31121874 on Build GRCh38

View Full Product Details